Merck
  • Antibiotic susceptibility profile of bifidobacteria as affected by oxgall, acid, and hydrogen peroxide stress.
Antibiotic susceptibility profile of bifidobacteria as affected by oxgall, acid, and hydrogen peroxide stress.
Antimicrobial agents and chemotherapy (2006-10-25)
E Kheadr, N Dabour, C Le Lay, C Lacroix, I Fliss
ABSTRACT
The effects of acid, oxgall, and H(2)O(2) on susceptibilities to antibiotics and nisin were examined for 13 strains of bifidobacteria. Susceptibilities to ampicillin, cloxacillin, penicillin, vancomycin, kanamycin, neomycin, paramomycin, streptomycin, chloramphenicol, erythromycin, tetracycline, and nisin A were assayed by a microdilution broth method. Acid-, oxgall- and H(2)O(2)-stressed variants were produced and assayed. Exposure to a pH of 2.0 for 60 min reduced susceptibilities to cloxacillin and nisin A but increased susceptibilities to ampicillin, vancomycin, aminoglycosides, chloramphenicol, and erythromycin in a strain-dependent manner. Exposure to oxgall (0.3%) for 90 min increased susceptibilities to cell wall-directed antibiotics and aminoglycosides but increased resistances to tetracycline and nisin A. Oxidative stress increased the susceptibilities of 70% of the strains to ampicillin and chloramphenicol, of 50% of the strains to cloxacillin and tetracycline, and of 40% of the strains to erythromycin but did not affect susceptibilities to vancomycin, kanamycin, and nisin A. This study shows that exposure of bifidobacteria to stressful conditions resembling those in the gastrointestinal tract may substantially modify their susceptibilities to antibiotics and may thus affect their probiotic capacities, especially when they are used for the management of intestinal infections and antibiotic-associated diarrhea.
Sigma-Aldrich
Cloxacillin sodium salt, ≥95.0% (HPLC)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture