Skip to Content
Merck
All Photos(4)

Key Documents

T7660

Sigma-Aldrich

Tetracycline hydrochloride

powder, BioReagent, suitable for cell culture

Synonym(s):

Tetracycline HCL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
Beilstein:
3844873
EC Number:
MDL number:
UNSPSC Code:
51284043
PubChem Substance ID:
NACRES:
NA.76

product line

BioReagent

Quality Level

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

yellow

mp

220-223 °C (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15-,21+,22-;/m0./s1

InChI key

XMEVHPAGJVLHIG-FMZCEJRJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Application

Tetracycline is a broad spectrum polyketide antibiotic with clinical uses in treating bacterial infections such as Rocky Mountain spotted fever, typush fever, tick fevers, Q fever, and Brill-Zinsser disease and to treat upper respiratory infections and acne. It has been used in studies of multidrug resistance and potential side effects including acute pancreatitis. It is recommended for use in cell culture applications at 10 mg/L.

Biochem/physiol Actions

Mode of Action: Tetracycline passively diffuses through proin channels in the cell membrane, binding to 30S ribosomes and inhibits protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. It also binds to the bacterial 50S ribosomal subunit, altering the membrane and causing intracellular components to leak from bacterial cells. The inhibitory effects can be reversed by washing, suggesting that it is the reversibly bound antibiotic, and not the irreversibly bound drug, that is responsible for antibacterial action.

Mode of Resistance: The effects are inactivated via a loss of cell wall permeability.

Antimicrobial spectrum: Includes a wide range of antimicrobial activity against gram-positive and gram-negative bacteria.

Caution

This product should be frozen below 0°C and protected from light and moisture. In these conditions, the product has been shown to retain activity for 4 years. Stock solutions should be stored at -20°C and are stable at 37°C for 4 days.

Preparation Note

The product is freely soluble in water, slightly soluble in 96% ethanol, and is practically insoluble in acetone. Tetracycline is rapidly destroyed by alkali hydroxide solutions and standing water solutions become turbid due to hydrolysis and precipitation. The potency of tetracycline is reduced in solutions with pH below 2.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place

Signal Word

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Emma Rossi et al.
Nature communications, 9(1), 4830-4830 (2018-11-18)
Dna2 is a DNA helicase-endonuclease mediating DSB resection and Okazaki fragment processing. Dna2 ablation is lethal and rescued by inactivation of Pif1, a helicase assisting Okazaki fragment maturation, Pol32, a DNA polymerase δ subunit, and Rad9, a DNA damage response
Marta Lukačišinová et al.
Nature communications, 11(1), 3105-3105 (2020-06-21)
Genetic perturbations that affect bacterial resistance to antibiotics have been characterized genome-wide, but how do such perturbations interact with subsequent evolutionary adaptation to the drug? Here, we show that strong epistasis between resistance mutations and systematically identified genes can be
Hana Antonicka et al.
Cell metabolism, 32(3), 479-497 (2020-09-03)
We used BioID, a proximity-dependent biotinylation assay with 100 mitochondrial baits from all mitochondrial sub-compartments, to create a high-resolution human mitochondrial proximity interaction network. We identified 1,465 proteins, producing 15,626 unique high-confidence proximity interactions. Of these, 528 proteins were previously
Michael L van de Weijer et al.
Molecular cell, 79(5), 768-781 (2020-08-02)
Misfolded proteins in the endoplasmic reticulum (ER) are degraded by ER-associated degradation (ERAD). Although ERAD components involved in degradation of luminal substrates are well characterized, much less is known about quality control of membrane proteins. Here, we analyzed the degradation
Sami Nourreddine et al.
Cell reports, 31(7), 107660-107660 (2020-05-21)
In human cells, the expression of ∼1,000 genes is modulated throughout the cell cycle. Although some of these genes are controlled by specific transcriptional programs, very little is known about their post-transcriptional regulation. Here, we analyze the expression signature associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service