Skip to Content
Merck
Sulfanilic acid JIS special grade, 99.0-100.5%

Sulfanilic acid

Sulfanilic acid JIS special grade, 99.0-100.5%
Synonym(s):
Sulfanilic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid
Linear Formula:
4-(H2N)C6H4SO3H
CAS Number:
Molecular Weight:
173.19
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique