
Oxytetracycline hydrochloride

Synonym(s):
5-Hydroxytetracycline hydrochloride, Oxytetracycline hydrochloride
Empirical Formula (Hill Notation):
C22H24N2O9 · HCl
CAS Number:
Molecular Weight:
496.89
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing