Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Safe-Lock microcentrifuge tubes
Products
Product Category
tubes (7)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Volume (ml)
Color
Package Size
Special Grade
Natural (1)
Sterility
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
T9661
Eppendorf
®
Safe-Lock microcentrifuge tubes,
volume 1.5 mL, natural
Expand
View Pricing
EP022363212
Eppendorf
®
Safe-Lock microcentrifuge tubes,
size 1.5 mL, PCR clean, colorless, pkg of 500 ea
Expand
View Pricing
EP0030123611
Eppendorf
®
Safe-Lock microcentrifuge tubes,
Forensic DNA Grade, volume 1.5 mL, pack of 500 tubes (10 bags x 50 pcs.)
Expand
View Pricing
EP022363344
Eppendorf
®
Safe-Lock microcentrifuge tubes,
PCR clean, volume 2.0 mL, colorless, pack of 500
Expand
View Pricing
EP0030123620
Eppendorf
®
Safe-Lock microcentrifuge tubes,
Forensic DNA Grade, volume 2.0 mL, pack of 500 tubes (10 bags x 50 pcs.)
Expand
View Pricing
EP022363719
Eppendorf
®
Safe-Lock microcentrifuge tubes,
size 0.5 mL, PCR clean, colorless, pkg of 500 ea
Expand
View Pricing
EP0030123603
Eppendorf
®
Safe-Lock microcentrifuge tubes,
Forensic DNA Grade, volume 0.5 mL, pack of 500 tubes (10 bags x 50 pcs.)
Expand
View Pricing