Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Atropine sulfate
Synonym(s):
Atropine sulfate salt monohydrate, α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, Tropine tropate
Linear Formula:
(C
17
H
23
NO
3
)
2
· H
2
SO
4
· H
2
O
CAS Number:
5908-99-6
Molecular Weight:
694.83
EC Number:
200-235-0
Products
Brand
Supelco (1)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Special Grade
pharmaceutical primary standard (3)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1045009
Atropine sulfate,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PHR1379
Atropine sulfate,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
A1400000
Atropine sulfate,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP023
Atropine sulfate,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing