Skip to Content
Merck
All Photos(1)

Key Documents

1607007

USP

Saccharin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,3-Dihydroxy-1,2-benzisothiazol-3-one-1,1-dioxide, 2-Sulfobenzoic acid imide, o-Benzoic sulfimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO3S
CAS Number:
Molecular Weight:
183.18
Beilstein:
6888
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

saccharin

manufacturer/tradename

USP

mp

226-229 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C1NS(=O)(=O)c2ccccc12

InChI

1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)

InChI key

CVHZOJJKTDOEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Saccharin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Saccharin Sodium Tablets

Biochem/physiol Actions

A sweet tastant for mammals. A glycerol taste receptor binding site specific for glucose has been proposed in drosophila.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Anderson
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 26(7), 637-644 (1988-07-01)
An hypothesis is presented of a mechanism for the sodium saccharin (NaS)-associated tumorigenesis of the urinary bladder that occurs in male rats. The ingestion of high doses of NaS is associated with increased urine volume and bladder mass. In rats
D L Arnold et al.
Toxicology, 27(3-4), 179-256 (1983-07-01)
Saccharin, first synthesized in 1879, eventually became popular as an inexpensive substitute for sugar, particularly as a non-caloric sweetner. The dispute concerning the safety of saccharin for human consumption is almost as old as saccharin itself. In this article, the
Marilyn E Carroll et al.
Behavioural pharmacology, 19(5-6), 435-460 (2008-08-12)
A positive relationship between the consumption of sweetened dietary substances (e.g. saccharin and sucrose) and drug abuse has been reported in both the human and other animal literature. The proposed genetic contribution to this relationship has been based on evidence
A G Renwick
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 23(4-5), 429-435 (1985-04-01)
Recent studies on saccharin in animals and man have allowed a detailed understanding of its fate in the body. Saccharin is slowly absorbed from the gut but rapidly eliminated in the urine, largely by renal tubular secretion. Saccharin does not
J Whysner et al.
Pharmacology & therapeutics, 71(1-2), 225-252 (1996-01-01)
Sodium saccharin (NaSac) produces bladder tumors consistently in male rats only after lifetime exposure that begins at birth. NaSac is not metabolized and is negative in most genotoxicity tests. NaSac-induced cell damage and proliferation have been proposed as important factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service