Skip to Content
Merck

1457083

USP

Naphthalene

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H8
CAS Number:
Molecular Weight:
128.17
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1421310
eCl@ss:
39011601
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

4.4 (vs air)

vapor pressure

0.03 mmHg ( 25 °C)

API family

naphthalene

form

flakes

autoignition temp.

978 °F

expl. lim.

5.9 %

manufacturer/tradename

USP

bp

218 °C (lit.)

mp

80-82 °C (lit.)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

c1ccc2ccccc2c1

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H

InChI key

UFWIBTONFRDIAS-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Naphthalene is a polycyclic aromatic hydrocarbon that is occasionally used in biologics formulations, particularly in processes where its role as a solvent or stabilizer is beneficial. In the biologics industry, naphthalene is valued for its ability to interact with certain bioactive molecules, helping to stabilize compounds in formulations and enhancing their efficacy in therapeutic applications.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Naphthalene USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Light Mineral Oil
  • Mineral Oil
  • Paraffin

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pha
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.


Still not finding the right product?

Explore all of our products under Naphthalene


signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3

flash_point_f

173.3 °F - closed cup

flash_point_c

78.5 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library