Skip to Content
Merck
All Photos(1)

Key Documents

1453005

USP

Naloxone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(5α)-4,5-Epoxy-3,14-dihydroxy-17-(2-propen-1-yl)morphinan-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

naloxone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N1([C@H]2[C@]3([C@@]4([C@@H](Oc5c4c(ccc5O)C2)C(=O)CC3)CC1)O)CC=C

InChI

1S/C19H21NO4/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11/h2-4,14,17,21,23H,1,5-10H2/t14-,17+,18+,19-/m1/s1

InChI key

UZHSEJADLWPNLE-GRGSLBFTSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Naloxone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Naloxone Hydrochloride
  • Naloxone Hydrochloride Injection
  • Pentazocine and Naloxone Tablets
  • Buprenorphine and Naloxone Sublingual Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naloxone Hydrochloride
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(2) (2022)
Pentazocine and Naloxone Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(1), 3463-3463 (2023)
Daisuke Yamada et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(8), 1852-1860 (2014-02-13)
Although the underlying mechanism remains unknown, several studies have suggested benefits of n-3 long-chain polyunsaturated fatty acid (PUFA) for patients with anxiety disorders. Elevated fear is thought to contribute to the pathogenesis of particular anxiety disorders. The aim of the
Marco Lanza et al.
British journal of pharmacology, 171(15), 3693-3701 (2014-04-25)
CR4056 is a novel imidazoline-2 (I2 ) ligand exhibiting potent analgesic activity in animal models of pain. In this study, we investigated the effects of CR4056 in a well-established model of postoperative pain where rats develop hyperalgesia in the injured
Nina Eriksen et al.
International journal of stroke : official journal of the International Stroke Society, 9(6), 802-810 (2012-08-30)
Strokes have both ischemic and hemorrhagic components, but most studies of experimental stroke only address the ischemic component. This is likely because investigations of hemorrhagic transformation are hindered by the lack of methods based on unbiased principles for volume estimation.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service