Skip to Content
Merck
All Photos(1)

Documents

1445470

USP

Mometasone furoate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(11β,16α)-9,21-Dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30Cl2O6
CAS Number:
Molecular Weight:
521.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mometasone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5ccco5)C(=O)CCl

InChI

1S/C27H30Cl2O6/c1-15-11-19-18-7-6-16-12-17(30)8-9-24(16,2)26(18,29)21(31)13-25(19,3)27(15,22(32)14-28)35-23(33)20-5-4-10-34-20/h4-5,8-10,12,15,18-19,21,31H,6-7,11,13-14H2,1-3H3/t15-,18+,19+,21+,24+,25+,26+,27+/m1/s1

InChI key

WOFMFGQZHJDGCX-ZULDAHANSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mometasone furoate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Mometasone furoate is an anti-inflammatory glucocorticoid.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Penagos et al.
Allergy, 63(10), 1280-1291 (2008-08-30)
Several randomized, double-blind, placebo-controlled clinical trials have demonstrated the efficacy of mometasone furoate nasal spray (MFNS) in the treatment of allergic rhinitis (AR) thus allowing for a meta-analysis to determine the overall treatment effect. A comprehensive search of the MEDLINE
Paul L McCormack et al.
Drugs, 66(8), 1151-1168 (2006-06-23)
Inhaled mometasone furoate (Asmanex) is a synthetic corticosteroid indicated for the first-line maintenance prophylactic therapy of persistent asthma in adults and adolescents. It is formulated for delivery via a breath-actuated dry powder inhaler (DPI) [Twisthaler].Inhaled mometasone furoate delivered by DPI
Michelle Zeidler et al.
Current medical research and opinion, 26(6), 1295-1305 (2010-04-08)
Inhaled corticosteroids (ICSs) are effective controller medications that treat the chronic inflammation of asthma. The goal of asthma treatment is to improve lung function, symptoms, and the ability to perform daily activities, while decreasing the risk of exacerbations. Mometasone furoate
C van Drunen et al.
Allergy, 60 Suppl 80, 5-19 (2005-06-14)
Mometasone furoate nasal spray (MFNS; Nasonex, Schering-Plough Corporation, Kenilworth, NJ, USA) is an effective and well-tolerated intranasal corticosteroid approved for the prophylactic treatment of seasonal allergic rhinitis, and the treatment of perennial allergic rhinitis. MFNS is a potent molecule with
I Haj-Salem et al.
Allergy, 70(2), 212-219 (2014-12-03)
Allergic asthma is characterized by inflammation and airway remodeling. Bronchial epithelium is considered a key player in coordinating airway wall remodeling. In mild asthma, the epithelium is damaged and fails to proliferate and to repair, whereas in severe asthma, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service