Skip to Content
Merck
All Photos(1)

Key Documents

1375502

USP

Mebendazole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-Benzoyl-2-benzimidazolecarbamic acid methyl ester, Mebendazol, Methyl N-(5-benzoyl-1H-benzimidazol-2-yl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13N3O3
CAS Number:
Molecular Weight:
295.29
Beilstein:
759809
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mebendazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccccc3

InChI

1S/C16H13N3O3/c1-22-16(21)19-15-17-12-8-7-11(9-13(12)18-15)14(20)10-5-3-2-4-6-10/h2-9H,1H3,(H2,17,18,19,21)

InChI key

OPXLLQIJSORQAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mebendazole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Mebendazole Tablets
  • Mebendazole Oral Suspension
  • Fenbendazole
  • Pimozide Tablets
  • Pimozide

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R W Ammann et al.
Journal of hepatology, 29(6), 994-998 (1999-01-06)
Long-term chemotherapy of human alveolar echinococcosis with benzimidazole compounds (mebendazole, albendazole) has been shown to be primarily parasitostatic, but its curative (parasitocidal) efficacy is debated. This article reports on a 67-year-old male patient with non-resectable alveolar echinococcosis of the liver
Mebendazole and related anthelmintics.
H Van den Bossche et al.
Advances in pharmacology and chemotherapy, 19, 67-128 (1982-01-01)
Olga Staudacher et al.
Tropical medicine & international health : TM & IH, 19(7), 812-824 (2014-04-23)
Preventive chemotherapy of schoolchildren against soil-transmitted helminths (STHs) is widely implemented in Rwanda. However, data on its actual efficacy are lacking. We assessed prevalence, associated factors and manifestation of STH infection among schoolchildren in southern highland Rwanda as well as
R Cañete et al.
Transactions of the Royal Society of Tropical Medicine and Hygiene, 103(5), 437-442 (2009-02-07)
Mebendazole, a benzimidazole carbamate compound, is currently in use for human medical practice against soil-transmitted helminthiasis (STH) and enterobiasis. However, it has been demonstrated that its spectrum of activity is broad and goes beyond those infections. Several studies provide evidence
A Bennett et al.
Parasitology today (Personal ed.), 16(2), 71-74 (2000-02-01)
The widespread use of mebendazole and albendazole for treating intestinal nematode infections in human populations is raising concerns that careful monitoring pro- cedures should be in place to identify any emergence of drug resistance. In this article, Andy Bennett and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service