1234668
USP
Eleutheroside B
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(E)-4-[-3-Hydroxy-1-propenyl]-2,6-dimethoxyphenyl-β-D-glucopyranoside, Ilexanthin A, Ligustrin, Lilacin, Magnolenin, Methoxyconiferine, Sinapyl alcohol 4-O-glucoside, Syringin, Syringoside
About This Item
grade
pharmaceutical primary standard
API family
eleutheroside
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
COc1cc(\C=C\CO)cc(OC)c1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O
InChI
1S/C17H24O9/c1-23-10-6-9(4-3-5-18)7-11(24-2)16(10)26-17-15(22)14(21)13(20)12(8-19)25-17/h3-4,6-7,12-15,17-22H,5,8H2,1-2H3/b4-3+/t12-,13-,14+,15-,17+/m1/s1
InChI key
QJVXKWHHAMZTBY-GCPOEHJPSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Eleuthero Root and Rhizome
- Eleuthero Root and Rhizome Dry Extract
- Eleuthero Root and Rhizome Dry Extract Capsules
- Eleuthero Root and Rhizome Dry Extract Tablets
- Eleuthero Root and Rhizome Powder
- Eleuthero Root and Rhizome Powder Capsules
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service