Skip to Content
Merck
All Photos(1)

Documents

1134368

USP

Citric acid

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

citric acid

expl. lim.

8 %, 65 °F

manufacturer/tradename

USP

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

153-159 °C (lit.)

solubility

water: 383 g/L at 25 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Citric acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Anhydrous Citric Acid
  • Citric Acid Monohydrate
  • Magnesium Carbonate, Citric Acid, and Potassium Citrate for Oral Solution
  • Monoglyceride Citrate
  • Powdered Garcinia Hydroxycitrate Extract
  • Anticoagulant Citrate Phosphate Dextrose Solution
  • Potassium Citrate Extended-Release Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnesium Carbonate, Citric Acid, and Potassium Citrate for Oral Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(3), 2711-2711 (2018)
Monoglyceride Citrate
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 30(6), 5891-5891 (2018)
Tricitrates Oral Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(3), 4504-4504 (2018)
Calcium Citrate Malate
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(2), 694-694 (2018)
Powdered Garcinia Hydroxycitrate Extract
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 36(3), 5014-5014 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service