Skip to Content
Merck
All Photos(1)

Key Documents

1011007

USP

Acitretin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acitretin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1cc(C)c(\C=C\C(C)=C\C=C\C(C)=C\C(O)=O)c(C)c1C

InChI

1S/C21H26O3/c1-14(8-7-9-15(2)12-21(22)23)10-11-19-16(3)13-20(24-6)18(5)17(19)4/h7-13H,1-6H3,(H,22,23)/b9-7+,11-10+,14-8+,15-12+

InChI key

IHUNBGSDBOWDMA-AQFIFDHZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acitretin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Acitretin Capsules

Biochem/physiol Actions

Synthetic retinoid that is a metabolite of etretinate. Preferentially binds to cellular retinoic acid binding proteins (CRABPs).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chai Sue Lee et al.
Expert opinion on drug safety, 8(6), 769-779 (2009-12-10)
Acitretin is an oral retinoid that is approved for the treatment of psoriasis. It is unique compared to other systemic therapies for psoriasis such as methotrexate and cyclosporine in that it is not immunosuppressive. It is, therefore, safe for use
Alessandro Borghi et al.
Acta dermato-venereologica, 95(3), 332-336 (2014-07-01)
The efficacy and safety of acitretin was evaluated retrospectively in a cohort of 46 patients with moderate to severe plaque psoriasis (Psoriasis Area and Severity Index (PASI) range 10-42). Patients were treated at an initial dose of 10 mg/day acitretin
Agranulocytosis and total scalp alopecia following acitretin.
T A Chave et al.
The British journal of dermatology, 148(5), 1063-1064 (2003-06-06)
British Association of Dermatologists guidelines on the efficacy and use of acitretin in dermatology.
A D Ormerod et al.
The British journal of dermatology, 162(5), 952-963 (2010-04-29)
Lauren K Dunn et al.
Journal of drugs in dermatology : JDD, 10(7), 772-782 (2011-07-02)
Acitretin is a systemic retinoid drug used in the treatment of severe psoriasis. It has also been used for a spectrum of other difficult-to-treat dermatoses, including hyperkeratotic and inflammatory dermatoses and non-melanoma skin cancers. Here we review the available data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service