Skip to Content
Merck
All Photos(1)

Documents

RAD170S

radiello Passive Sampler Starter Kit

Hydrogen Sulfide

Synonym(s):

radiello Starter Kits

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

product name

radiello Hydrogen Sulfide Starter Kit,

Agency

NIOSH 6013
OSHA 1008

Quality Level

packaging

pkg of 1 kit

manufacturer/tradename

Radiello

technique(s)

passive air sampling: suitable

matrix

microporous PE coated with zinc acetate

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling Hydrogen sulfide

Looking for similar products? Visit Product Comparison Guide

General description

Radiello Starter Kits are ideal for introducing you to the product and include everything you need to take one complete sample. No additional parts are required. Each kit includes the following items:

Each RAD130S Starter Kit Includes:
Two radiello cartridge adsorbents - Solvent Desorption Note: 1 x cartridge for sampling; and 1 x for blank)
1 x Triangular support plate (RAD121)
1 x vertical adapter for personal sampling (RAD122)
1 x white diffusive body (RAD120)
1 x barcode label (RAD190)
Detailed sampling/analysis instructions


Each RAD141S and 145S Starter Kit Includes:
Two radiello cartridge adsorbents - Thermal Desorption (Note: 1 x cartridge for sampling; and 1 x for blank)
1 x Triangular support plate (RAD121)
1 x vertical adapter for personal sampling (RAD122)
1 x yellow diffusive body (RAD1202)
1 x barcode label (RAD190)
Detailed sampling/analysis instructions

Packaging

Kit includes: 2 RAD170 adsorbing cartridges and barcode labels, 1 RAD120 white diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet

Legal Information

radiello is a trademark of Institi Clinici Scientifici Maugeri

Disclaimer

*Note: This product is not officially specified in the method, it may be suitable for use for sampling compound(s) listed in the method*

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
Armelle Vallat et al.
Phytochemistry, 66(13), 1540-1550 (2005-06-14)
Headspace volatiles from apple-bearing twigs were collected in the field with a Radiello sampler during three different diurnal periods over the complete fruit growing season. Analyses by thermal desorption-GC-MS identified a total of 62 compounds in changing quantities, including the
Francesc A Esteve-Turrillas et al.
Journal of chromatography. A, 1216(48), 8549-8556 (2009-10-27)
A new procedure for the passive sampling in air of benzene, toluene, ethylbenzene and xylene isomers (BTEX) is proposed. A low-density polyethylene layflat tube filled with a mixture of solid phases provided a high versatility tool for the sampling of
Joelle Roukos et al.
Environmental pollution (Barking, Essex : 1987), 157(11), 3001-3009 (2009-07-08)
Two measurement campaigns of volatile organic compounds (VOC) were carried out in the industrial city of Dunkerque, using Radiello passive samplers during winter (16-23 January) and summer (6-13 June) 2007. 174 compounds were identified belonging to six chemical families. Classifying
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic

Articles

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Protocols

Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.

Related Content

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service