Skip to Content
Merck
All Photos(1)

Key Documents

ERMFC440B

Resin-bonded glass fibre board (thermal conductivity)

ERM®, certified reference material, size 50 cm × 50 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

size

50 cm × 50 cm

application(s)

glass & ceramic

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please click on the below product number:
ERMFC440B

Legal Information

ERM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service