Skip to Content
Merck
All Photos(1)

Key Documents

8S61336

Supelco

SS Benzoic Acid

2000 μg/mL in dichloromethane, analytical standard

Synonym(s):

Benzoic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Agency

EPA 8270

Assay

99.90%

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 2.5 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

This product is being deleted. Product PHR1050-1G may be an acceptable replacement.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Nacht et al.
Journal of the American Academy of Dermatology, 4(1), 31-37 (1981-01-01)
The transepidermal penetration and metabolic disposition of 14C-benzoyl peroxide were assessed in vitro (excised human skin) and in vivo (rhesus monkey). In vitro, the benzoyl peroxide penetrated into the skin, through the stratum corneum or the follicular openings, or both
G C Tremblay et al.
Pharmacology & therapeutics, 60(1), 63-90 (1993-10-01)
Detoxification of sodium benzoate by elimination as a conjugate with glycine, a nonessential amino acid, provides a pathway for the disposal of waste nitrogen. Since 1979, sodium benzoate has been widely used in the therapeutic regimen to combat ammonia toxicity
Renbo Wei et al.
Macromolecular rapid communications, 34(4), 330-334 (2013-01-03)
A new approach is developed to fabricate highly oriented mono-domain LCE nano/microstructures through micro-molding in capillaries. Gratings and microwires as two typical examples are fabricated and characterized by polarizing optical microscopy, optical microscopy, and scanning electron microscopy. The gratings with
Dajie Lin et al.
Biosensors & bioelectronics, 45, 195-200 (2013-03-19)
An amplification strategy for signal tracing was developed by introducing a host-guest binding reaction into the assembly process of gold nanorods (AuNRs) superstructure. The amplification pathway firstly used a thio-β-cyclodextrin (SH-β-CD) functionalized gold nanoparticles to label signal antibody, and then
Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service