Skip to Content
Merck
All Photos(1)

Key Documents

821960

TSKgel® Q-STAT (10 µm) HPLC Column

L x I.D. 3.5 cm × 3 mm, ammonium ion, quaternary phase

Synonym(s):

TSKgel® Q-STAT Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501
L × I.D.:
3.5 cm × 3 mm
particle size:
10 μm
matrix active group:
ammonium ion, quaternary phase
matrix:
polymer particle platform

Product Name

TSKgel® Q-STAT HPLC Column, phase ammonium ion, quaternary, L × I.D. 3.5 cm × 3 mm, 10 μm particle size

material

stainless steel hardware

Agency

suitable for USP L23

product line

TSKgel®

feature

counter ion Cl-

manufacturer/tradename

Tosoh Q-STAT, 21960

parameter

no limitation salt concentration
≤50% organic solvent
1.0-2.0 mL/min standard flow rate
10-60 °C temperature
100 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

3.5 cm × 3 mm

matrix

polymer particle platform

matrix active group

ammonium ion, quaternary phase

particle size

10 μm

operating pH

3-10

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel Q-STAT columns are packed with 7 and 10μm hydrophilic non-porous resin particles of which the surface consists of an open access network of multi-layered anion exchange groups. The innovative bonding chemistry, combined with a relatively large particle size, results in a respectable loading capacity and a low operating pressure, making these columns suitable for all HPLC and FPLC systems. Applications for the TSKgel Q-STAT columns include the separation of: proteins, peptides, low molecular weight nucleic acids, aggregates and charge isomers of monoclonal antibodies, PEGylated proteins, oligo DNA, and siRNA.

Physical form

Shipped in DI water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

FPLC is a trademark of Cytiva
TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service