Skip to Content
Merck
All Photos(1)

Key Documents

59564C30

Supelco

SUPELCOSIL LC-18 (5 µm) HPLC Columns

L × I.D. 2 cm × 3 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm

material

stainless steel column

Agency

suitable for USP L1

product line

SUPELCOSIL

feature

endcapped

packaging

pkg of 2 ea

parameter

<70 °C temp. range

technique(s)

HPLC: suitable

L × I.D.

2 cm × 3 mm

surface area

170 m2/g

matrix

silica gel high purity, spherical
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

pH range

2-7.5

application(s)

food and beverages

compatibility

use to protect LC-18, LC-PAH

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K M Matar et al.
Therapeutic drug monitoring, 21(5), 559-566 (1999-10-16)
A simple, rapid, sensitive, and reproducible high-performance liquid chromatographic (HPLC) method for simultaneous determination of the antiepileptic drugs (ethosuximide, primidone, lamotrigine, phenobarbital, phenytoin, and carbamazepine) and two metabolites (carbamazepine-diol and carbamazepine-epoxide) in human plasma is described. The procedure involves extraction
O Cudina et al.
Farmaco (Societa chimica italiana : 1989), 55(2), 125-127 (2000-04-27)
Fluocortolone and its esters are synthetic corticosteroids used topically in the treatment of various skin disorders. A method that can be successfully used for the separation and determination of fluocortolone pivalate and fluocortolone hexanoate in suppositories was developed. This method
V Stocchi et al.
Analytical biochemistry, 167(1), 181-190 (1987-11-15)
A simple and fast ion pair reversed-phase high-performance liquid chromatographic method has been developed for the simultaneous determination of ATP, ADP, AMP, GTP, GDP, IMP, NADP+, NADPH+, NAD+, NADH, ADP-ribose, inosine, adenosine, hypoxanthine, and xanthine. This method allows us to
G R Granneman et al.
Clinical pharmacokinetics, 29 Suppl 2, 1-8 (1995-01-01)
A rapid and sensitive assay was developed for the measurement of plasma concentrations of zileuton racemate, a potent inhibitor of 5-lipoxygenase. Zileuton and its inactive N-dehydroxylated metabolite were extracted from human, monkey, and rat plasma by use of a solid-phase
L Marai et al.
Canadian journal of biochemistry and cell biology = Revue canadienne de biochimie et biologie cellulaire, 61(8), 840-849 (1983-08-01)
Natural triacylglycerols were resolved by high pressure liquid chromatography on Supelcosil LC-18 columns using a gradient of 30-90% propionitrile in acetonitrile as eluting solvent. The effluent was admitted to a quadrupole mass spectrometer via a direct liquid inlet interface. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service