Key Documents
58686
Tube Fitting, Flangeless
1/4-28 male Upchurch nut and ferrule for 1/8 in. O.D. tubing
About This Item
UNSPSC Code:
41121815
material
Delrin® nut
Tefzel® ferrule (flangeless)
packaging
pkg of 5 ea
parameter
500 psi max. pressure (35 bar)
fitting
1/4-28 male Upchurch nut and ferrule for 1/8 in. O.D. tubing
General description
Tube Fitting, Flangeless can be used as a replacement part for Supelco mobile phase reservoir system.
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)