Skip to Content
Merck
All Photos(1)

Key Documents

51164-U

Supelco

EXP® Pre-Column Filter Cartridges

0.5μm, pkg of 5 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.

packaging

pkg of 5 ea

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

EXP is a registered trademark of Optimize Technologies, Inc.

required but not provided

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

BIOshell™ IgG 1000 Å C4 UHPLC Column for Improved Biomacromolecule Separations

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service