Skip to Content
Merck
All Photos(1)

Key Documents

49453-U

Supelco

C4 - C24 Even Carbon Saturated FAMEs

1000 μg/mL each component in hexane, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

storage temp.

2-8°C

General description

Quantitative standard suitable for use as a retention index marker probe.

Check out our complete portfolio of Fatty acid methyl ester (FAME) reference material solution mixes

Application

Essential Oil Analysis Essential oils are complex mixtures of chemical compounds produced by living organisms and isolated from specific plant parts of known taxonomic origin. Their composition mainly includes monoterpenes, sesquiterpenes, and their oxygenated derivatives, as well as aldehydes, alcohols, and esters. In collaboration with Chromaleont and Shimadzu, our integrated solutions enhance efficiency and sustainability for the analysis of essential oils, addressing demanding workflows through innovative research, case studies, consumables, and applications.

Analyte

Description

    Methyl arachidate
    Methyl behenate
    Methyl butyrate
    Methyl decanoate
    Methyl hexanoate
    Methyl dodecanoate
    Methyl myristate
    Methyl octanoate
    Methyl palmitate
    Methyl stearate
    Methyl tetracosanoate
    See All (11)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Fappi et al.
BioMed research international, 2014, 961438-961438 (2014-07-02)
Corticosteroids cause muscle atrophy by acting on proteasomal and lysosomal systems and by affecting pathways related to muscular trophysm, such as the IGF-1/PI-3k/Akt/mTOR. Omega-3 fatty acid (n-3) has been used beneficially to attenuate muscle atrophy linked to sepsis and cachexia;
José M Bastías et al.
PloS one, 12(7), e0180993-e0180993 (2017-07-08)
The effect of four cooking methods was evaluated for proximate composition, fatty acid, calcium, iron, and zinc content in salmon and Chilean jack mackerel. The moisture content of steamed salmon decreased (64.94%) compared to the control (68.05%); a significant decrease
María Gallardo et al.
Animals : an open access journal from MDPI, 9(2) (2019-02-20)
This experiment was carried out to determine the effect of breed on mRNA and protein expression levels of lipogenic enzymes acetyl-CoA carboxylase α (ACC), fatty acid synthase (FAS), stearoyl-CoA desaturase 1 (SCD1) plus sterol regulatory element binding transcription factor 1c
Gabriela Grassmann Roschel et al.
Journal of food science, 86(2), 411-419 (2021-01-16)
Echium seed oil is an alternative source of omega 3 fatty acids but it is highly susceptible to oxidation. A combination of three natural strategies was proposed in this study aiming to improve the oxidative stability of echium oil obtained
Rávila Graziany Machado de Souza et al.
Nutrition (Burbank, Los Angeles County, Calif.), 55-56, 154-160 (2018-08-08)
Nut consumption is associated with reduced risks of cardiovascular disease. Baru almonds have a high protein content and high quantities of mono- and polyunsaturated fatty acids, phenolic compounds, and antioxidants. This study aimed to evaluate the effects of a baru

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service