Skip to Content
Merck
All Photos(1)

Documents

48873

Supelco

ASTM® D2887 Reference Gas Oil

pkg of 6 × 1 mL

Synonym(s):

RGO

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Agency

ASTM® D2887,D2887,D7169

CofA

current certificate can be downloaded

packaging

pkg of 6 × 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

ASTM ® D2887 reference gas oil can be used as standard in order to measure the boiling range of petroleum fractions, like diesel fuel and gasoline with gas chromatography method(GC).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 1B - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Speight.GJ
Environmental Analysis and Technology for the Refining Industry (2005)
Song C
Chemistry of Diesel Fuels (2000)

Articles

Boiling point range distribution analysis is crucial for petrochemical product processing, control, contaminant identification, and method development.

Boiling point range distribution analysis is crucial for petrochemical product processing, control, contaminant identification, and method development.

Boiling point range distribution analysis is crucial for petrochemical product processing, control, contaminant identification, and method development.

Boiling point range distribution analysis is crucial for petrochemical product processing, control, contaminant identification, and method development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service