Skip to Content
Merck
All Photos(1)

Key Documents

47786

Supelco

DL-alpha-Tocopherol acetate

analytical standard

Synonym(s):

Vitamin E acetate, O-Acetyl-α-tocopherol, DL-α-Tocopherol acetate, DL-α-Tocopheryl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
Beilstein:
97512
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

feature

standard type fat soluble vitamin

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.496 (lit.)

bp

224 °C/0.3 mmHg (lit.)

density

0.953 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

-10 to -25°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(C)=O)c(C)c(C)c2O1

InChI

1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1

InChI key

ZAKOWWREFLAJOT-CEFNRUSXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takafumi Matsumura et al.
Scientific reports, 11(1), 3458-3458 (2021-02-12)
In vitro spermatogenesis (IVS) using air-liquid interphase organ culture method is possible with mouse testis tissues. The same method, however, has been hardly applicable to animals other than mice, only producing no or limited progression of spermatogenesis. In the present
Kristin I Løken-Amsrud et al.
PloS one, 8(1), e54417-e54417 (2013-01-26)
Alpha-tocopherol is the main vitamin E compound in humans, and has important antioxidative and immunomodulatory properties. The aim of this study was to study alpha-tocopherol concentrations and their relationship to disease activity in Norwegian multiple sclerosis (MS) patients. Prospective cohort
Katharina M Espe et al.
Clinical journal of the American Society of Nephrology : CJASN, 8(3), 452-458 (2013-01-22)
Trials with the antioxidant vitamin E have failed to show benefit in the general population. Considering the different causes of death in ESRD, this study investigated the association between plasma concentrations of α-tocopherol and specific clinical outcomes in diabetic hemodialysis
Po-Hsien Huang et al.
Science signaling, 6(267), ra19-ra19 (2013-03-21)
Vitamin E is a fat-soluble vitamin with antioxidant properties. Tocopherols are the predominant form of vitamin E found in the diet and in supplements and have garnered interest for their potential cancer therapeutic and preventive effects, such as the dephosphorylation
Chuan Liu et al.
Toxicology letters, 218(1), 2-9 (2013-01-22)
Whether exposure to radiofrequency electromagnetic radiation (RF-EMR) emitted from mobile phones can induce DNA damage in male germ cells remains unclear. In this study, we conducted a 24h intermittent exposure (5 min on and 10 min off) of a mouse

Protocols

GB method with Ascentis® Express C30 column achieves baseline resolution for Vitamin A and E critical pair.

GB method with Ascentis® Express C30 column achieves baseline resolution for Vitamin A and E critical pair.

GB method with Ascentis® Express C30 column achieves baseline resolution for Vitamin A and E critical pair.

GB method with Ascentis® Express C30 column achieves baseline resolution for Vitamin A and E critical pair.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service