Skip to Content
Merck
All Photos(1)

Documents

47773

Supelco

Phylloquinone (K1)

analytical standard

Synonym(s):

Vitamin K1, 2-Methyl-3-phytyl-1,4-naphthoquinone, 3-Phytylmenadione, Phylloquinone, Vitamin K1(20)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H46O2
CAS Number:
Molecular Weight:
450.70
Beilstein:
2568816
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

Quality Level

form

oil

CofA

current certificate can be downloaded

feature

standard type fat soluble vitamin

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.527 (lit.)

mp

−20 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC=C21)C(C)=C(C/C=C(CCC[C@@H](CCC[C@H](C)CCCC(C)C)C)\C)C2=O

InChI

1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+/t23-,24-/m1/s1

InChI key

MBWXNTAXLNYFJB-NKFFZRIASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phylloquinone (K1) is a prenylated naphthoquinone. It is a fat soluble vitamin, produced by plants, green algae and some species of cyanobacteria. It functions as a coenzyme and is important for the synthesis of a number of proteins involved in blood clotting and bone metabolism.

Application

Phylloquinone (K1) may be used as an analytical reference standard for the quantification of the analyte in olive oil, chard, human plasma samples, animal and plant food products using reversed-phase high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of a simplified HPLC assay for the determination of phylloquinone (vitamin K1) in animal and plant food items
Jakob E and Elmadfa I
Food Chemistry, 56(1), 87-91 (1996)
Determination of vitamin K1 content in olive oil, chard and human plasma by RP-HPLC method with UV?Vis detection
Otles S and Cagindi O
Food Chemistry, 100(3), 1220-1222 (2007)
Xi Gu et al.
Plant physiology, 185(4), 1443-1456 (2021-04-02)
Nonphotosynthetic holoparasites exploit flexible targeting of phylloquinone biosynthesis to facilitate plasma membrane redox signaling. Phylloquinone is a lipophilic naphthoquinone found predominantly in chloroplasts and best known for its function in photosystem I electron transport and disulfide bridge formation of photosystem
Rogier Caluwé et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 29(7), 1385-1390 (2013-11-29)
Haemodialysis patients suffer from accelerated vascular calcification. The vitamin K-dependent matrix Gla protein (MGP) is one of the most powerful inhibitors of vascular calcification. Haemodialysis patients have high levels of the inactive form of MGP (desphosphorylated-uncarboxylated-MGP, dp-uc-MGP) and may benefit
Ellen C M Cranenburg et al.
Kidney international, 82(5), 605-610 (2012-06-01)
Vitamin K is essential for the activity of γ-carboxyglutamate (Gla)-proteins including matrix Gla28 protein and osteocalcin; an inhibitor of vascular calcification and a bone matrix protein, respectively. Insufficient vitamin K intake leads to the production of non-carboxylated, inactive proteins and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service