Skip to Content
Merck
All Photos(1)

Key Documents

47287-U

Supelco

Lactose Monohydrate

analytical standard

Synonym(s):

α-Lactose monohydrate, β-D-Gal-(1→4)-α-D-Glc, 4-O-β-D-Galactopyranosyl-α-D-glucose, Milk sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

analyte chemical class(es)

oligosaccharides

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-30°C

SMILES string

[H]O[H].OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11.H2O/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17;/h3-20H,1-2H2;1H2/t3-,4-,5+,6+,7-,8-,9-,10-,11+,12+;/m1./s1

InChI key

WSVLPVUVIUVCRA-KPKNDVKVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ibrahima Youm et al.
Journal of pharmaceutical sciences, 105(10), 3162-3171 (2016-07-28)
It is hypothesized that ferrocene (FC)-loaded nanocarriers (FC-NCs) are safe label-free contrast agents for cochlear biodistribution study by transmission electron microscopy (TEM). To test this hypothesis, after engineering, the poly(epsilon-caprolactone)/polyglycolide NCs are tested for stability with various types and ratios
Eduardo Costa Pinto et al.
Pharmaceutics, 4(3), 430-441 (2012-01-01)
The purpose of this study was to propose an analytical procedure that provides the effects of particle size and surface area on dissolution of efavirenz. Five different batches obtained by different micronization processes and with different particle size distribution and
Hannah L Keizer et al.
International journal of pharmaceutics, 573, 118810-118810 (2019-11-05)
Roll compaction/dry granulation is a widely used granulation method in the pharmaceutical industry. The simulation of the process is of great interest, especially in the early phase of formulation development of solid dosage forms. The hybrid modeling approach allows to
Mai Tal et al.
ACS photonics, 7(12), 3286-3290 (2020-12-29)
Nonlinear plasmonic metasurfaces provide new and promising means to produce broadband terahertz (THz) radiation, due to their compact size and functionalities beyond those achievable with conventional THz emitters. However, they were driven to date only by amplified laser systems, which
Amy E Rohlfing et al.
PLoS genetics, 15(7), e1008224-e1008224 (2019-07-06)
The gastrointestinal pathogen, Clostridioides difficile, initiates infection when its metabolically dormant spore form germinates in the mammalian gut. While most spore-forming bacteria use transmembrane germinant receptors to sense nutrient germinants, C. difficile is thought to use the soluble pseudoprotease, CspC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service