Skip to Content
Merck
All Photos(1)

Documents

47268-U

Supelco

Disaccharides Kit

(individually packaged in quantities indicated), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

analyte chemical class(es)

oligosaccharides

packaging

pkg of 1 kit

concentration

(individually packaged in quantities indicated)

technique(s)

GC/MS: suitable
HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-30°C

General description

Disaccharides Kit contains individually packaged standards in the quantities listed. The disaccharides are prepared, tested, and individually packaged using rigorous manufacturing procedures.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Kit Components Only

Product No.
Description

  • Isomaltose (mixed anomers) 100 mg

  • α-Lactose 500 mg

  • Maltose (47288) 500 mg

  • Sucrose (47289) 500 mg

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Zhang et al.
Asian-Australasian journal of animal sciences, 27(6), 855-861 (2014-07-23)
This experiment was performed to investigate the effects of exogenous xylanase supplementation on performance, nutrient digestibility and the degradation of non-starch polysaccharides (NSP) in different sections of the gastrointestinal tract (GIT) of broilers fed wheat-based diets. A total of 120

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service