Skip to Content
Merck
All Photos(1)

Key Documents

442543

Supelco

Decafluorotriphenylphosphine

analytical standard

Synonym(s):

Bis(pentafluorophenyl)phenylphosphine, DFTPP, 2,3,4,5,6,2′,3′,4′,5′,6′-Decafluorotriphenylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5P(C6F5)2
CAS Number:
Molecular Weight:
442.19
Beilstein:
6253897
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

55-60 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Fc1c(F)c(F)c(P(c2ccccc2)c3c(F)c(F)c(F)c(F)c3F)c(F)c1F

InChI

1S/C18H5F10P/c19-7-9(21)13(25)17(14(26)10(7)22)29(6-4-2-1-3-5-6)18-15(27)11(23)8(20)12(24)16(18)28/h1-5H

InChI key

OYNXPGGNQMSMTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decafluorotriphenylphosphine (DFTPP) is commonly used as a check standard for the calibration of mass spectrum detector (MSD) prior to the analysis of samples under investigation.

Application

DFTPP may be used as a reference compound to assess the abundance performance/calibration of gas chromatography-mass spectrometry (GC-MS) systems prior to the determination of polychlorinated biphenyls in human serum, pesticides in whole blood analysis, nonsteroidal anti-inflammatory drugs (NSAIDs) in blood samples of clinical and forensic cases, organic priority pollutants in the composts of sewage sludge with rice straw and micropollutants in environmental and food samples.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and reliable screening method for detection of 70 pesticides in whole blood by gas chromatography-mass spectrometry using a constructed calibration-locking database.
Kudo K, et al.
Legal medicine, 14(2), 93-100 (2012)
Rapid screening of 18 nonsteroidal anti-inflammatory drugs (NSAIDs) using novel [TM="NAGINATA"] gas chromatography-mass spectrometry software
Nagamatsu K, et al.
Forensic Toxicology, 30(1), 11-18 (2012)
Novel gas chromatography-mass spectrometry database for automatic identification and quantification of micropollutants
Kadokami K, et al.
Journal of Chromatography A, 1089(1-2), 219-226 (2005)
Analysis of polychlorinated biphenyls in human serum by gas chromatography-mass selective detection operating at high ion source temperature.
Tran BN, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 877(11-12), 1109-1116 (2009)
Identifying the potential sources of di-(2-ethylhexyl) phthalate contamination in the sediment of the Houjing River in southern Taiwan.
Lin C, et al.
Journal of Hazardous Materials, 161(1), 270-275 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service