Skip to Content
Merck
All Photos(1)

Documents

24205-U

Supelco

VOCOL® Capillary GC Column

L × I.D. 30 m × 0.25 mm, df 1.50 μm

Synonym(s):

FS CAP VOCOL® 30M .25MM 1.5UM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
eCl@ss:
32119290
NACRES:
SB.54

material

fused silica

Quality Level

Agency

EPA TO-17,502.2,624,8015,8260,OLM04.2 VOA,524.2
NIOSH 1003
suitable for EPA 601

parameter

≤25-250 °C temperature (isothermal or programmed)

Beta value

42

df

1.50 μm

technique(s)

gas chromatography (GC): suitable

L × I.D.

30 m × 0.25 mm

application(s)

environmental
food and beverages
forensics and toxicology
industrial hygiene

column type

capillary intermediate polar

Looking for similar products? Visit Product Comparison Guide

General description

Application: This intermediate polarity column, designed for analyses of volatile organic compounds (VOCs), offers great retention and resolution of highly volatile compounds. Use this column in direct injection ports or coupled to purge and trap systems.
USP Code: None
Phase:
  • Bonded
  • Proprietary
Temp. Limits:
  • ≤0.32 mm I.D., <2 μm: Subambient to 250 °C (isothermal or programmed)
  • ≤0.32 mm I.D., ≥2 μm: Subambient to 230 °C (isothermal or programmed)
  • ≥0.53 mm I.D., <2 μm: Subambient to 250 °C (isothermal or programmed)
  • ≥0.53 mm I.D., ≥2 μm: Subambient to 230 °C (isothermal or programmed)

Application

VOCOL® column (diphenyl dimethyl polysiloxane with crosslinking moieties) may be used for their relevancy to the purge-and-trap technique and GC/MS analysis of 34 volatile organics. It was also found suitable in being used for determination of volatile compounds by whole column cryotrapping.

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

VOCOL is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of volatile compounds in water by purging directly to a capillary column with whole column cryotrapping.
Pankow JF and Rosen ME.
Environmental Science & Technology, 22 (4), 398-405 (1988)
Nathalie Henrich et al.
Logopedics, phoniatrics, vocology, 32(4), 171-177 (2007-11-09)
Are the characteristic timbre and loudness of Bulgarian women's singing related to tuning of resonances of the vocal tract? We studied an Australian female singer, who practises and teaches Bulgarian singing technique. Two different vocal qualities of this style were
Carmelle Moore
Logopedics, phoniatrics, vocology, 37(4), 144-150 (2012-11-10)
This paper explores the application of modified yoga techniques, as an adjunct to voice therapy, by a speech pathologist who is also a yoga teacher. Yoga practices, with effects that may be short-term, are not considered a substitute for comprehensive
Katherine Sanchez et al.
Logopedics, phoniatrics, vocology, 39(2), 62-71 (2013-04-18)
Automatic recordings were made of speech and voice range profiles for 63 vocally healthy Australian men and women without voice training (30 males and 33 females aged 21 to 65 years). Test-retest reliability, evaluated for a subgroup, was high. Speech
Irena Vincent et al.
Logopedics, phoniatrics, vocology, 37(1), 22-32 (2011-12-14)
To investigate voice changes as they develop over time due to cigarette smoking, women who never smoked (NS), women who smoked less than 10 years (S1), and women who smoked 10 or more years (S2) were compared. Acoustic (fundamental frequency

Articles

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

HS-SPME with Carboxen®/PDMS coated Nitinol fiber allows low MW analyte detection in light-exposed milk plastics.

Protocols

In this article, SPME-GC was successfully used to identify many volatile compounds present in the headspace over several spirits.

There are three types of analyses: GRO, DRO, and TPH. The published methods are simply variations of these to meet the different analytical needs of the individual agencies.

There are three types of analyses: GRO, DRO, and TPH. The published methods are simply variations of these to meet the different analytical needs of the individual agencies.

There are three types of analyses: GRO, DRO, and TPH. The published methods are simply variations of these to meet the different analytical needs of the individual agencies.

See All

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Chromatograms

suitable for GCsuitable for GC, application for SPMEsuitable for GC, application for SPMEsuitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service