Key Documents
21993
Supelco
VOST Stack Sampling Tube
greener alternative
empty
About This Item
UNSPSC Code:
41104008
Pricing and availability is not currently available.
Quality Level
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
greener alternative category
General description
  • VOST Tubes (Volatile Organic Sampling Train) are designed to meet specifications in US EPA SW-846, Method 0030.
  • Each tube is individually numbered, pre-conditioned, and sealed with stainless steel Swagelok fittings before stored in a glass storage container.
  • Each lot is tested for background and backpressure.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)