Skip to Content
Merck
All Photos(1)

Key Documents

21130-U

Supelco

Supelpak-2 polymeric adsorbent, XAD-2

matrix styrene-divinylbenzene, 20-60 mesh, jar of 1000g

Synonym(s):

XAD®-2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

Product Name

Supelpak-2, matrix styrene-divinylbenzene, 20-60 mesh, jar of 1000 g

description

purified XAD-2

form

powder

packaging

jar of 1000 g

parameter

200 °C max. temp.

technique(s)

GC/MS: suitable
LPLC: suitable

surface area

~300 m2/g

matrix

styrene-divinylbenzene

matrix active group

polymer

particle size

20-60 mesh

pore size

~0.65 mL/g pore volume
90 Å mean pore size

density

1.02 g/mL at 25 °C (true wet)(lit.)
1.07 g/mL at 25 °C (skeletal)(lit.)

application(s)

environmental

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Supelpak-2 is purified Amberlite XAD®-2 that has been cleaned to meet and exceed US EPA-recommended criteria for purity, as outlined in Level I Environmental Assessment Procedures Manual. It is the best resin to use for standard air sampling methods requiring resin tested for background TCO (total chromatographic organics) level. Packaged in glass containers.

Application

Supelpak-2 was used as adsorbent in extraction procedure and these extracts were analysed using GC-MS, GC-FTIR, proton NMR and GC-NPD

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
Supelpak is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F J Egea González et al.
Journal of AOAC International, 80(5), 1091-1097 (1997-11-05)
A method to sample and analyze chlorothalonil and dichlofluanid in greenhouse air was evaluated. Analysis was performed by gas chromatography with electron capture detection and gas chromatography-mass spectrometry. Solid sorbents such as Chromosorb 102, Porapak R, Supelpak-2, Amberlite XAD-2, Amberlite
Olivier Briand et al.
The Science of the total environment, 288(3), 199-213 (2002-05-07)
An analytical method using gas chromatography-mass spectrometry has been developed for the evaluation of different sampling techniques to characterise spray drift in a commercial apple orchard. Eleven pesticides were studied (fungicides, insecticides and herbicides). A collection of airborne spray-drift pesticides
F J Egea González et al.
Journal of chromatography. A, 829(1-2), 251-258 (1999-01-29)
The diminution of metamidophos residue levels with time in vegetables an greenhouse air was investigated after treatment of tomatoes and green beans. A gas chromatographic method using dichloromethane as an extraction solvent has been developed to analyse metamidophos in vegetables
D Rochat et al.
Journal of chemical ecology, 17(11), 2127-2141 (1991-11-01)
Male American palm weevils (APWs),Rhynchophorus palmarum (L.) produced two sex-specific compounds, which were disclosed by volatile collections on Supelpak-2 and gas chromatography. One was a minor compound, not always detected. The major male-produced volatile was identified as (2E)-6-methyl-2-hepten-4-ol through coupled
Nikolaos G Tsiropoulos et al.
Analytica chimica acta, 573-574, 209-215 (2007-08-29)
A methodology is described for greenhouse air analysis by sampling fenhexamid, pyrimethanil, malathion, metalaxyl-M and myclobutanil in solid sorbents. Pesticides were determined by gas chromatography with NP Detector. The trapping efficiency of XAD-2, XAD-4, Supelpak-2, Florisil and C-18 at different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service