Skip to Content
Merck
All Photos(1)

Documents

20305

Supelco

Molecular Sieve, 13Å

60-80 mesh, bottle of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
eCl@ss:
32110204

product name

Molecular Sieve Adsorbent, matrix Molecular Sieve 13X, 60-80 mesh, bottle of 50 g

form

solid

Quality Level

packaging

bottle of 50 g

matrix

Molecular Sieve 13X

particle size

60-80 mesh

pore size

13 Å pore diameter

density

~0.65 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

Molecular sieves are synthetically produced zeolites (naturally occurring aluminosilicate minerals), and are characterized by pores and internal cavities of extremely uniform dimensions. These crystalline materials have three-dimensional structures based on silicon oxide (SiO4) and aluminum oxide (AlO4) polyhedra. The polyhedra are linked by their corners to produce an open structure with internal cavities in which molecules can be trapped. These materials are engineered so that access to the internal cavities is through specific and uniform sized pores.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of siloxanes in biogases.
Schweigkofler M and Niessner R.
Journal of Hazardous Materials, 183-196 (2001)
Columns for Gas Chromatography: Performance and Selection.
Barry EF and Grob RL.
Science, 28-28 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service