Skip to Content
Merck
All Photos(1)

Key Documents

20033

Supelco

ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg

W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea

Synonym(s):

ORBO Activated Silica Gel Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900
Pricing and availability is not currently available.

material

W,W,W separators
silica gel

Quality Level

Agency

NIOSH 2012

product line

ORBO

composition

Bed A, 150 mg
Bed B, 75 mg

packaging

pkg of 50 ea

manufacturer/tradename

ORBO 554

technique(s)

active air sampling: suitable

O.D. × L

6 mm × 75 mm

matrix

H2SO4 coated silica gel

particle size

20-40 mesh

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

  • ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg solvent desorption tube is designed for gas and vapor sampling.
  • It contains two beds of the same selective adsorbent separated by glass wool or foam.
  • Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.

Legal Information

ORBO is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Orbo et al.
International journal of oncology, 5(3), 619-625 (1994-09-01)
About 50% of the cases with invasive carcinoma of the uterine cervix occurs before menopause. Cyclic changes of sex steroids may therefore interfere with the ratio between extracellular levels of cGMP and cAMP (cGMP(ex)/cAMP(ex)). This ratio has been proposed as
H Stalsberg et al.
Virchows Archiv. A, Pathological anatomy and histopathology, 414(5), 415-422 (1989-01-01)
The autopsies of 12 victims from two snow avalanches in North-Norway are reported. Supportive evidence from non-autopsied and surviving victims is included. Consistent autopsy findings were prominent lung oedema, moderate cerebral oedema, extreme contraction of the left ventricle, petechiae in
Anne Ørbo et al.
The Journal of steroid biochemistry and molecular biology, 113(1-2), 139-149 (2009-01-14)
Patients with endometrial hyperplasia representing preliminary stages of endometrial cancer have shown to respond to therapy in 100% of the cases when treated with levonorgestrel-impregnated intrauterine device. Anti-proliferative effect has also been reported after application of an anti-progestin impregnated intrauterine
Anne Beate Vereide et al.
Gynecologic oncology, 97(3), 740-750 (2005-05-12)
The aim of the study was to investigate apoptosis as a growth regulatory mechanism of gestagen in endometrial precancers and to compare differences in the apoptotic cascade after high and low dose gestagen regimens. Pre- and post-treatment paraffin-embedded endometrial hyperplasia
A Ørbo et al.
BJOG : an international journal of obstetrics and gynaecology, 123(9), 1512-1519 (2015-12-03)
To investigate relapse rates after the successful treatment of patients with non-atypical endometrial hyperplasia who were randomised to either a levonorgestrel-impregnated intrauterine system (LNG-IUS; Mirena(®) ) or two regimens of oral medroxyprogesterone acetate (MPA) after primary histological response. A multicentre

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service