Skip to Content
Merck
All Photos(18)

Key Documents

ZRB1869

Sigma-Aldrich

Anti-HNF-3A/FOXA1 Antibody, clone 1E5 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
FACS
ICC
IHC (p)
WB
affinity binding assay
clone:
1E5, recombinant monoclonal
species reactivity:
human
citations:
technique(s):
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E5, recombinant monoclonal

description

1E5 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 49.15 kDa
observed mol wt ~52 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FOXA1(3169)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E5 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Hepatocyte nuclear factor 3a (HNF-3A/FOXA1). It targets an epitope within 14 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of human Hepatocyte nuclear factor 3a (HNF-3A/FOXA1).

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected HNF-3A/FOXA1 in HepG2 cell lysate.

Tested applications

Flow Cytometry Analysis: 0.1 μg from a representative lot detected HNF-3A/FOXA1 in one million HepG2 cells.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected HNF-3A/FOXA1 in HepG2 and MCF-7 cells.

Affinity Binding Assay: A representative lot of this antibody bound HNF-3A/FOXA1 with a KD of 1.0 x 10-12 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected HNF-3A/FOXA1 in human prostate tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Hepatocyte nuclear factor 3-alpha (UniProt: P55317; also known as HNF-3-alpha, HNF-3A, Forkhead box protein A1, Transcription factor 3A, TCF-3A) is encoded by the FOXA1 (also known as HNF3A, TCF3A) gene (Gene ID: 3169) in human. HNF-3A is a transcription factor that is involved in embryonic development, establishment of tissue-specific gene expression, and regulation of gene expression in differentiated tissues. It acts as a ′pioneer′ factor opening the compacted chromatin for other proteins through interactions with nucleosomal core histones and thereby replacing linker histones at target enhancer and/or promoter sites. It is also involved in cell cycle regulation by activating expression of p27Kip1 (CDKN1B), alone or in conjunction with BRCA1. In healthy adults, it is expressed in gut endoderm-derived tissues, particularly in the liver. However, high expression of HNF-3A is reported in prostate and estrogen receptor 1-positive breast tumors and in esophageal and lung adenocarcinomas. It binds DNA with the consensus sequence 5′-[AC]A[AT]T[AG]TT[GT][AG][CT]T[CT]-3′ and its DNA binding region is localized in amino acids 169-260 (Forkhead). This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Williamson, EA., et al. (2006). Oncogene. 25(9); 1391-1399; Lai, E., et al. (1993). Proc. Natl. Acad. Sci. USA. 90 (22); 10421-10423).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service