ZRB1861
Anti-Bcl-xL Antibody, clone 1O8 ZooMAb® Rabbit Monoclonal
recombinant, expressed in HEK 293 cells
Synonym(s):
Apoptosis regulator Bcl-X, Bcl-2-like protein 1, Bcl2-L-1
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1O8, recombinant monoclonal
description
1O8 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 26.05 kDa
observed mol wt ~28 kDa
purified by
using Protein A
species reactivity
monkey, human, mouse
species reactivity (predicted by homology)
canine, bovine, porcine, feline, rat
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
technique(s)
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... BCL2L1(598)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in COS-1 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Bcl-xL in COS-1 cell lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Bcl-xL in MCF-7 and RAW264.7 cell lysates and 1:10,000 dilution detected the recombinant active form of Human Bcl-xL.
Flow Cytometry Analysis: 0.1 µg from a representative lot detected Bcl-xL in one million MCF-7 cells.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Bcl-xL in MCF-7 cells.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Bcl-xL in human bone marrow tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service