Key Documents
Z768618
Nalgene® square graduated media bottle
without cap, PET, sterile, capacity 1000 mL
Synonym(s):
media storage bottle, square plastic bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
capacity:
1000 mL
material:
PET
square bottle
sterility:
sterile
feature:
graduations
without cap
packaging:
pack of 24 ea (2 x pack of 12)
material
PET
square bottle
Joint
Joints threaded neck (38-430)
sterility
sterile
feature
graduations
without cap
packaging
pack of 24 ea (2 x pack of 12)
manufacturer/tradename
Nalgene® 342044-1000 (USA)
Nalgene® 842044-1000 (European)
capacity
1000 mL
General description
Nalgene® Square PET media bottles offer sterility, lightweight construction, and shatter resistance. They are specifically designed for use in filling lines.
Features and Benefits
  • Has molded-in graduations and neck-ring.
  • Non-cytotoxic, non-hemolytic, and non-pyrogenic.
  • Exhibits exceptional gas barrier properties.
  • Gamma irradiated to 10^-6 to achieve sterility assurance level (SAL).
  • Composed of material free from ADC (animal derived component).
Other Notes
Closures for these bottles are sold separately.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)