Z741975
GVS Transfer Membrane
Nitrocellulose, 0.45 um, W × L 30 cm × 3 m, roll of 1 roll
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
nitrocellulose membrane
sterility
non-sterile
packaging
roll of 1 roll
manufacturer/tradename
GVS Life Sciences 1215471
W × L
30 cm × 3 m
pore size
0.45 μm
General description
Description and Use
GVS Pure Nitrocellulose Transfer Membrane is the membrane of choice for all protein or immunoblotting applications.The high sensitivity of GVS Pure Nitrocellulose Transfer Membrane ensures excellent results in all transfers, especially in protein blotting.
Features & Benefits
Typical Applications
GVS Pure Nitrocellulose Transfer Membrane is the membrane of choice for all protein or immunoblotting applications.The high sensitivity of GVS Pure Nitrocellulose Transfer Membrane ensures excellent results in all transfers, especially in protein blotting.
Features & Benefits
- For procedures that require optimum resolution
- Membrane of choice for protein or immunoblotting applications
- Low background, easily blocked
- BSA binding capacity up to 100μg/cm2
- Wets out naturally
- Compatible with all detection systems
Typical Applications
- Westerns
- Protein & immunoblotting
- Northerns
- Southerns
- Dot/slot blots
- Radiographic, chromogenic and chemiluminescent detection systems
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of immunology (Baltimore, Md. : 1950), 204(6), 1598-1606 (2020-02-12)
C1q/TNF-related protein (CTRP) 6 is a member of the CTRP protein family associated with the regulation of cellular and endocrine processes. CTRP6 contains collagen and globular structures, resembling the pattern recognition molecules (PRMs) of the classical and lectin complement pathways.
Journal of Cancer, 11(8), 2318-2328 (2020-03-05)
Introduction: For pathological diagnosis of pancreatic neuroendocrine neoplasms (pNENs) the routinely used immunohistochemical markers are chromogranin A (CgA) and synaptophysin (Syn). Their ability as prognostic markers is not well established. A splice variant of actinin-4 (Actn-4sv) was recently found to
Molecular and cellular endocrinology, 503, 110696-110696 (2020-01-01)
We aim to investigate the expression of the glucagon receptor (GCGR) in human adipose tissue, and the impact of glucagon in glucose uptake and lipolysis in human adipocytes. GCGR gene expression in human subcutaneous and visceral adipose tissue was demonstrated
Thoracic cancer, 11(3), 537-548 (2020-01-10)
Circular RNAs (circRNAs) participate in the development of human cancers by regulating multiple cell processes. CircRNA antisense to the cerebellar degeneration-related protein 1 transcript (circCDR1as) expression is dysregulated in many cancers, including non-small-cell lung cancer (NSCLC). However, the mechanism by
Neuroscience letters, 715, 134558-134558 (2019-10-23)
Periostin has dual roles as a multifunctional cytokine and an extracellular matrix protein that binds to several integrins on a variety of cells. This study evaluated whether the expression and cellular localization of periostin changes in the spinal cord of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service