Skip to Content
Merck
All Photos(1)

Key Documents

Z4277

Sigma-Aldrich

Zileuton

≥98% (HPLC)

Synonym(s):

(±)-N-hydroxy-N-(1-benzo[b]thien-2-ylethyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2S
CAS Number:
Molecular Weight:
236.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL at ~60 °C (warm up to 60 C for 5min)

originator

Abbott

storage temp.

room temp

SMILES string

CC(N(O)C(N)=O)c1cc2ccccc2s1

InChI

1S/C11H12N2O2S/c1-7(13(15)11(12)14)10-6-8-4-2-3-5-9(8)16-10/h2-7,15H,1H3,(H2,12,14)

InChI key

MWLSOWXNZPKENC-UHFFFAOYSA-N

Gene Information

human ... ALOX5(240)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Zileuton is a 5-Lipoxygenase inhibitor.
Zileuton is an anti-asthmatic, an inhibitor of 5-lipoxygenase; the initial enzyme in the biosynthesis of leukotrienes from arachidonic acid.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norwin Kubick et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-10)
Repurposing drugs to target M1 macrophages inflammatory response in depression constitutes a bright alternative for commonly used antidepressants. Depression is a significant type of mood disorder, where patients suffer from pathological disturbances associated with a proinflammatory M1 macrophage phenotype. Presently
Hee Young Kim et al.
eLife, 12 (2024-07-09)
Trained immunity is the long-term functional reprogramming of innate immune cells, which results in altered responses toward a secondary challenge. Despite indoxyl sulfate (IS) being a potent stimulus associated with chronic kidney disease (CKD)-related inflammation, its impact on trained immunity
Lalit Pukhrambam Singh et al.
JOJ ophthalmology, 8(5), 77-85 (2022-02-22)
Diabetic retinopathy (DR) is a devastating disease leading to blindness among majority of working adults around the globe. Nonetheless, an effective treatment or cure for the disease is still to be achieved. This is because the cellular and molecular mechanisms
Kaneez Fatima et al.
Frontiers in nutrition, 9, 857256-857256 (2022-04-26)
Carvacrol, a monoterpene known for its pharmacological activities, is present in the essential oil of Origanum majorana, Origanum vulgare, Thymus vulgaris, and Lippia graveolens. It is used in food as a flavoring and preservative agent in cosmetics and medicines because
Ju Youn Lee et al.
Nature communications, 11(1), 2358-2358 (2020-05-14)
Sphingosine kinase1 (SphK1) is an acetyl-CoA dependent acetyltransferase which acts on cyclooxygenase2 (COX2) in neurons in a model of Alzheimer's disease (AD). However, the mechanism underlying this activity was unexplored. Here we show that N-acetyl sphingosine (N-AS) is first generated

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service