Skip to Content
Merck
All Photos(1)

Key Documents

Z3777

Sigma-Aldrich

ZD7288 hydrate

≥98% (HPLC)

Synonym(s):

4-(N-Ethyl-N-phenylamino)-1,2 dimethyl-6-(methylamino) pyrimidinium chloride, ICI-D7288, N-Ethyl-1,6-dihydro-1,2-dimethyl-6-(methylimino)-N-phenyl-4-pyrimidinamine hydrochloride, ZD 7288

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20N4 · HCl · xH2O
Molecular Weight:
292.81 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

O.Cl.CCN(c1ccccc1)C2=CC(=N/C)\N(C)C(C)=N2

InChI

1S/C15H20N4.ClH.H2O/c1-5-19(13-9-7-6-8-10-13)15-11-14(16-3)18(4)12(2)17-15;;/h6-11H,5H2,1-4H3;1H;1H2/b16-14+;;

InChI key

NCEDDDWQLCMZQG-UPONXUSQSA-N

Application

ZD7288 hydrate has been used as Ih blocker to study its effect on
neocortical tissue. It has been used as hyperpolarization-activated and cyclic nucleotide-gated (HCN)-channel blocker to test the involvement of HCN channels in the phototransduction pathway.

Biochem/physiol Actions

Selective blocker of cation channel Ih; If blocker and sino-atrial node function modulator, blocks hyperpolarization-activated and cyclic nucleotide-gated (HCN) channels.
Selective blocker of cation channel Ih; If blocker and sino-atrial node function modulator, blocks hyperpolarization-activated and cyclic nucleotide-gated (HCN) channels.
ZD7288 can block hippocampal synaptic plasticity.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Cyclic Nucleotide-Gated (CNG) and Hyperpolarization Activated Cyclic Nucleotide-Gated (HCN) Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclic-Nucleotide-and HCN-Channel-Mediated Phototransduction in Intrinsically Photosensitive Retinal Ganglion Cells
Jiang Z, et al.
Cell, 175(3), 652-664 (2018)
Yingchao Xue et al.
Stem cells translational medicine, 8(2), 112-123 (2018-11-06)
Proneural transcription factors (TFs) drive highly efficient differentiation of pluripotent stem cells to lineage-specific neurons. However, current strategies mainly rely on genome-integrating viruses. Here, we used synthetic mRNAs coding two proneural TFs (Atoh1 and Ngn2) to differentiate induced pluripotent stem
The anticonvulsant lamotrigine enhances Ih in layer 2/3 neocortical pyramidal neurons of patients with pharmacoresistant epilepsy
Lehnhoff J, et al.
Neuropharmacology, 144, 58-69 (2019)
Marcin Siwiec et al.
Neuropharmacology, 177, 108248-108248 (2020-08-01)
Accumulating evidence suggests a widespread role of serotonin 5-HT7 receptors (5-HT7Rs) in the physiology of cognitive and affective processing. However, we still lack insights into 5-HT7R electrophysiology. Studies analyzing the 5-HT7R-mediated changes in CA1 pyramidal neuron activity revealed that 5-HT7R
Janna Lehnhoff et al.
Neuropharmacology, 144, 58-69 (2018-10-14)
Alterations of the hyperpolarization activated nonselective cation current (Ih) are associated with epileptogenesis. Accordingly, the second-generation antiepileptic drug lamotrigine (LTG) enhances Ih in rodent hippocampus. We directly evaluated here whether LTG fails to enhance Ih in neocortical slices from patients

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service