Skip to Content
Merck
All Photos(3)

Key Documents

WH0004904M1

Sigma-Aldrich

Monoclonal Anti-YBX1 antibody produced in mouse

clone 4F12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BP8, Anti-CSDB, Anti-DBPB, Anti-MDRNF1, Anti-MGC104858, Anti-MGC110976, Anti-NSEP1, Anti-YB1, Anti-YBX1, Anti-nuclease sensitive element binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YBX1(4904)

Related Categories

General description

Y-box binding protein 1 (YBX1) gene codes for Y-box protein 1 (YB-1) that has 324 amino acid residues, 8 exons and 7 introns. It is a member of the family of multifunctional DNA/RNA binding proteins. It is mainly present in the cytosol. YBX1 gene is mapped to human chromosome 1p34.

Immunogen

YBX1 (NP_004550, 51 a.a. ~ 139 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DKKVIATKVLGTVKWFNVRNGYGFINRNDTKEDVFVHQTAIKKNNPRKYLRSVGDGETVEFDVVEGEKGAEAANVTGPGGVPVQGSKYA

Biochem/physiol Actions

Y-box binding protein 1 (YBX1) participates in pre-mRNA splicing, transcriptional regulation and mRNA translation and stability. It is also involved in DNA repair and environmental stress responses and chromatin remodeling.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

YBX1 (Y box binding protein 1)
Evdokimova V and Sorokin A
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2011)
Andreas Zaucker et al.
Nucleic acids research, 46(1), 104-119 (2017-10-24)
In many organisms, transcriptional and post-transcriptional regulation of components of pathways or processes has been reported. However, to date, there are few reports of translational co-regulation of multiple components of a developmental signaling pathway. Here, we show that an RNA
Jiawei Sun et al.
Development (Cambridge, England), 145(19) (2018-08-24)
Maternal mRNAs and proteins dictate early embryonic development before zygotic genome activation. In the absence of transcription, elaborate control of maternal mRNA translation is of particular importance for oocyte maturation and early embryogenesis. By analyzing zebrafish ybx1 mutants with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service