Skip to Content
Merck
All Photos(2)

Key Documents

WH0003596M1

Sigma-Aldrich

Monoclonal Anti-IL13 antibody produced in mouse

clone 7E4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ALRH, Anti-IL13, Anti-MGC116786, Anti-MGC116788, Anti-MGC116789, Anti-P600, Anti-interleukin 13

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7E4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL13(3596)

General description

This gene encodes an immunoregulatory cytokine produced primarily by activated Th2 cells. This cytokine is involved in several stages of B-cell maturation and differentiation. It up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. This cytokine down-regulates macrophage activity, thereby inhibits the production of pro-inflammatory cytokines and chemokines. This cytokine is found to be critical to the pathogenesis of allergen-induced asthma but operates through mechanisms independent of IgE and eosinophils. This gene, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with this gene particularly close to IL4. [provided by RefSeq

Immunogen

IL13 (NP_002179, a.a. 35-146) full length recombinant protein with GST tag. MW of the GST tag alone is 26 kDa.

Biochem/physiol Actions

Interleukin-13 (IL-13) has a role in immune activities against infections and influences the function of various cells taking part in the same. It has been studied to be overexpressed in glioblastoma tumors and cell lines. IL-13 stimulates fibrosis in many infectious diseases. Targeted deletion of IL-13 in mice resulted in impaired T-helper 2 (Th2) cell development and indicated an important role for IL-13 in the expulsion of gastrointestinal parasites. IL-13 exerts anti-inflammatory effects on monocytes and macrophages and it inhibits the expression of inflammatory cytokines such as tumor necrosis factor-α (TNF-α), interleukins-1β, -6 and -8. IL-13 has also been shown to enhance B cell proliferation and to induce isotype switching resulting in increased production of immunoglobulin E (IgE). Blocking of IL-13 activity inhibits the pathophysiology of asthma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-4, IL-10 and IL-13 down-regulate monocyte-chemoattracting protein-1 (MCP-1) production in activated intestinal epithelial cells.
Kucharzik T
Clinical and Experimental Immunology, 111(1), 152-157 (1998)
IL-13 induces proliferation, Ig isotype switching, and Ig synthesis by immature human fetal B cells.
Punnonen J and de Vries JE
Journal of Immunology, 152(3), 1094-1102 (1994)
T cell responses: naive to memory and everything in between.
Pennock ND
Advances in Physiology Education, 37(4), 273-283 (2013)
Qi Liu et al.
The Journal of heart valve disease, 24(4), 496-501 (2016-02-24)
Interleukin (IL)-13 is a major inducer of fibrosis in many chronic infectious diseases, yet few studies have reported its role in valvular fibrosis in patients with rheumatic heart disease (RHD). The study aim was to investigate the role of IL-13
IL-13 mediates IL-33-dependent mast cell and type 2 innate lymphoid cell effects on bronchial epithelial cells.
Deepti R Nagarkar et al.
The Journal of allergy and clinical immunology, 136(1), 202-205 (2015-03-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service