Skip to Content
Merck
All Photos(1)

Documents

V9130

Sigma-Aldrich

N-Vanillylnonanamide

≥97%, powder

Synonym(s):

Capsaicin synthetic, N-(4-Hydroxy-3-methoxybenzyl)nonanamide, N-Vanillylpelargonamide, Nonanoic acid vanillylamide, Nonylvanylamide, Pelargonic acid vanillylamide, Pseudocapsaicin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO3
CAS Number:
Molecular Weight:
293.40
Beilstein:
2144300
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97%

form

powder

color

white to off-white

solubility

methanol: 100 mg/mL, clear to slightly hazy, colorless to deep brown-yellow

storage temp.

2-8°C

SMILES string

CCCCCCCCC(=O)NCc1ccc(O)c(OC)c1

InChI

1S/C17H27NO3/c1-3-4-5-6-7-8-9-17(20)18-13-14-10-11-15(19)16(12-14)21-2/h10-12,19H,3-9,13H2,1-2H3,(H,18,20)

InChI key

RGOVYLWUIBMPGK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

N-Vanillylnonanamide has antifouling properties. It acts as an nicotinamide adenine dinucleotide phosphate (NADPH) inhibitor.

Application

N-Vanillylnonanamide has been used for the preparation of spicules. It has also been used to reduce the firing of most extrinsic sensory axons.

Other Notes

Synthetic analogue of capsaicin with similar bioactivity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of strains experienced by viscerofugal nerve cell bodies during mechanosensitive firing using digital image correlation
Palmer G, et al.
American Journal of Physiology. Heart and Circulatory Physiology (2016)
Sphingolipids in Disease (2013)
Federica Villa et al.
Biotechnology letters, 31(9), 1407-1413 (2009-06-03)
The potential on N-vanillylnonanamide (NVN) in preventing the attachment of Pseudomonas stutzeri and a Bacillus cereus-group strain was investigated. NVN up to 852 microM was not toxic, nor was it an energy source for either organism. Microbial attachment assays were
P A Durant et al.
Brain research, 451(1-2), 301-308 (1988-06-07)
Unanesthetized rats chronically implanted with vesical and intrathecal catheters were injected intrathecally (i.t.) with either capsaicin (CAP), N-vanillylnonanamide (VN), 6-hydroxydopamine (6-OHDA), or 5,6-dihydroxytryptamine (5,6-DHT). The volume-evoked micturition reflex was assessed by cystometrography before, and 2 h, 1 day and 7
S M Gardiner et al.
The American journal of physiology, 257(6 Pt 2), R1429-R1435 (1989-12-01)
The vasopressin-mediated recovery of arterial pressure observed in adult rats following pharmacological blockade of the sympathetic nervous and renin-angiotensin systems is reduced by neonatal capsaicin treatment. We now demonstrate a similar but reversible effect following treatment of adult Wistar or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service