Skip to Content
Merck
All Photos(6)

Key Documents

V2258

Sigma-Aldrich

Monoclonal Anti-Vimentin antibody produced in mouse

clone LN-6, ascites fluid

Synonym(s):

Anti-F5H288

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

LN-6, monoclonal

contains

15 mM sodium azide

species reactivity

pig, mouse, sheep, human, bovine, rabbit, feline, rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200 using human tissue sections
immunoprecipitation (IP): suitable
indirect immunofluorescence: suitable using cultured cells
western blot: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)
mouse ... Vim(22352)
rat ... Vim(81818)

General description

Vimentin belongs to the class of type III intermediate filament protein family. The gene is located on human chromosome 10p13. It is a cytoskeletal protein. Vimentin is mainly expressed in normal mesenchymal cells.
Vimentin is a 58 kD class III intermediate filament present mainly in mesenchymal cells. It regulates cell shape, motility and endurance of mechanical stress in mesenchymal cells. Monoclonal anti-vimentin antibody can be used for the localization of vimentin by various immunochemical assays like immunoblotting, immunohistochemistry and immunoprecipitation. Frozen B5 fixed and formalin fixed, paraffin embedded tissue sections can be stained using anti-vimentin antibody. Mouse monoclonal anti-vimentin antibody reacts specifically with an epitope of the cytoskeletal protein vimentin (58kD). The product shows cross reactivity with vimentin from bovine, human, pig, sheep, cat, rat, mouse and rabbit.

Specificity

Recognizes an epitope of the cytoskeleton protein vimentin that is not detected in human lymphoid cells or derived malignancies. It is therefore useful in differentiating sarcomas from lymphomas.

Immunogen

human thymic nuclear extract

Application

Monoclonal Anti-Vimentin antibody can be used in immunocytochemistry and immunohistochemical staining. It can also be used in immunofluorescence staining (diluted 1: 500) .
Monoclonal Anti-Vimentin antibody produced in mouse has been used in western blotting and immunolabeling.

Biochem/physiol Actions

Vimentin plays an important role in modulating cell growth. It plays a pivotal role in epithelial-mesenchymal transition. Vimentin acts as a potential target for anti-metastasis therapy for nasopharyngeal carcinoma (NPC). It regulates cellular architecture and tissue integrity. It is also involved in tumorigenesis and the metastatic spread of cancer. Vimentin provides stress resistance. It might be involved in neuritogenesis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margit A Huber et al.
The Journal of clinical investigation, 114(4), 569-581 (2004-08-18)
The transcription factor NF-kappaB is activated in a range of human cancers and is thought to promote tumorigenesis, mainly due to its ability to protect transformed cells from apoptosis. To investigate the role of NF-kappaB in epithelial plasticity and metastasis
Role of vimentin in early stages of neuritogenesis in cultured hippocampal neurons
Boyne LJ, et al.
International Journal of Developmental Neuroscience, 14(6), 739-748 (1996)
M J Gibson et al.
Endocrinology, 141(9), 3065-3071 (2000-08-31)
Axons of GnRH neurons terminate at the median eminence in the medial basal hypothalamus (MBH) of the brain early in development. Similarly, GnRH neurons in grafts of preoptic area (POA) tissue within the third ventricle of hypogonadal mice preferentially innervate
Vimentin is a crucial target for anti-metastasis therapy of nasopharyngeal carcinoma
Wang W, et al.
Molecular and Cellular Biochemistry, 438(1-2), 47-57 (2018)
Fractional CO2 laser: a novel therapeutic device upon photobiomodulation of tissue remodeling and cytokine pathway of tissue repair
Prignano F, et al.
Dermatologic Therapy, 22, S8-S15 (2009)

Articles

In the midst of beeping lab timers, presentations and grant deadlines, it is easy to take for granted the quality of lab reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service