Skip to Content
Merck
All Photos(1)

Key Documents

V0389

Sigma-Aldrich

Anti-VGLUT1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-BNPI, Anti-Brain-specific Na(+)-dependent inorganic phosphate cotransporter, Anti-Vesicular Glutamate Transporter 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60 kDa

species reactivity

human

technique(s)

microarray: suitable
western blot (chemiluminescent): 1:1,000 using synaptic vesicle (SV) fraction of rat brain.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC17A7(57030)

General description

VGLUT1 (vesicular glutamate transporter 1, originally termed BNPI and characterized as a Na+/Pi transporter) is a 60 kDa protein. VGLUT2 is enriched in synaptic vesicles and selective for a distinct class of glutaminergic nerve terminals.
Vesicular glutamate transporter 1 (VGLUT1), also called as solute carrier family 17 member 7 (SLC17A7), is encoded by the gene mapped to human chromosome 19q13, a region associated with schizophrenia. VGLUT1 is specifically expressed in neuronal cells in the brain and at higher levels in neuron-enriched regions such as the amygdala and hippocampus. Glia-enriched areas such as the corpus callosum also show moderate level of expression and substantia nigra, subthalamic nuclei and thalamus show low level expression. VGLUT1 is characterized with a conserved C-terminal dileucine-like motif and two polyproline domains distal to C- terminal end, including one that binds to endocytic BAR (Bin/Amphiphysin/Rvs) domain protein, endophilin.

Immunogen

synthetic peptide corresponding to a region near the C-terminus of human VGLUT1 (amino aicds 504-520).

Biochem/physiol Actions

Vesicular glutamate transporter 1 (VGLUT1) is associated with neuronal pathways that exhibit activity-dependent potentiation, whereas VGLUT2 is expressed in sensory and autonomic pathways that display high fidelity neurotransmission.
Vesicular glutamate transporter 1 (VGLUT1) plays a vital role as a vesicular glutamate transporter in glutamatergic neurons. VGLUT1 expression in the prefrontal cortex functions as a potential neurochemical marker of dementia in Alzheimer′s disease (AD) and antidepressant activity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VGLUTs in peripheral neurons and the spinal cord: time for a review
Brumovsky PR
ISRN neurology, 2013 (2013)
Alireza Kashani et al.
Neurobiology of aging, 29(11), 1619-1630 (2007-05-29)
Several lines of evidence suggest that the glutamatergic system is severely impaired in Alzheimer disease (AD). Here, we assessed the status of glutamatergic terminals in AD using the first available specific markers, the vesicular glutamate transporters VGLUT1 and VGLUT2. We
L Bai et al.
The Journal of biological chemistry, 276(39), 36764-36769 (2001-07-04)
Glutamate is the major excitatory neurotransmitter in the mammalian central nervous system. Packaging and storage of glutamate into glutamatergic neuronal vesicles requires ATP-dependent vesicular glutamate uptake systems, which utilize the electrochemical proton gradient as a driving force. VGLUT1, the first
Larissa Moutsimilli et al.
Neuropharmacology, 49(6), 890-900 (2005-08-23)
The two recently characterized vesicular glutamate transporters (VGLUT) presynaptically mark and differentiate two distinct excitatory neuronal populations and thus define a cortical and a subcortical glutamatergic system (VGLUT1 and VGLUT2 positive, respectively). These two systems might be differentially implicated in
Expression of vesicular glutamate transporters type 1 and 2 in sensory and autonomic neurons innervating the mouse colorectum
Brumovsky PR, et al.
The Journal of Comparative Neurology, 519(16), 3346-3366 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service