Skip to Content
Merck
All Photos(1)

Documents

U6628

Sigma-Aldrich

Uvaol

≥95%

Synonym(s):

Urs-12-ene-3,28-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O2
CAS Number:
Molecular Weight:
442.72
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95%

mp

223-225 °C (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

C[C@@H]1CC[C@]2(CO)CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C

InChI

1S/C30H50O2/c1-19-10-15-30(18-31)17-16-28(6)21(25(30)20(19)2)8-9-23-27(5)13-12-24(32)26(3,4)22(27)11-14-29(23,28)7/h8,19-20,22-25,31-32H,9-18H2,1-7H3/t19-,20+,22+,23-,24+,25+,27+,28-,29-,30-/m1/s1

InChI key

XUARCIYIVXVTAE-ZAPOICBTSA-N

Gene Information

mouse ... Nos2(18126)

Looking for similar products? Visit Product Comparison Guide

General description

Uvaolis a pentacyclic triterpene compound usually extracted from plant leaves such as Apocynum venetum L., olive leaves (Olea europaea L.), and oregano (Origanum vulgare L.).

Application

Uvaol has been used as retinoic acid receptor-related orphan receptor gamma t (RORγt) inhibitor to study the role of cardiac glycoside reductase 2 (Cgr2) enzyme on its metabolism.

Biochem/physiol Actions

Uvaolexerts pharmacological properties such as antioxidant, anticancer, anti-inflammatory, and wound healing. It also displays vasodilator, hepatoprotective, and antimicrobial effects. Uvaol has inhibitory effects on nitric oxide (NO) release.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Z Xu et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 25(4), 225-226 (2003-01-07)
To study the chemical components of Gentiana tizuensis distributed in Qinghai Province. Chromatography and spectral analyses were used to isolate the constituents and elucidate their structure. Four compounds were isolated and identified as uvaol, L-(+)-l-O-methyl-myo-inositol, ursolic acid,beta-sitosterol. All these components
L I Somova et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 11(2-3), 121-129 (2004-04-09)
The cardiotonic and antidysrhythmic effects of four triterpenoid derivatives, namely oleanolic acid (OA), ursolic acid (UA), and uvaol (UV), isolated from the leaves of African wild olive (Olea europaea, subsp. africana) as well as methyl maslinate (MM) isolated from the
A Martins et al.
Anticancer research, 30(3), 829-835 (2010-04-16)
A bioassay-guided separation protocol, including the testing of the extracts, fractions and pure compounds for their ability to inhibit P-glycoprotein (the efflux pump responsible for the multidrug resistance of the used cell line) of mouse lymphoma cells containing the human
De-Hong Huang et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(3), 375-378 (2011-08-10)
To study the chemical constituents of Coleus forskohlii. Isolation and purification were carried out by silica gel column chromatographic and Toyopearl HW-40F. Compounds were identified and elucidated by spectral and chemical methods. Seven compounds were obtained from ethyl acetate extract
Margaret Alexander et al.
Cell host & microbe, 30(1), 17-30 (2021-11-26)
Bacterial activation of T helper 17 (Th17) cells exacerbates mouse models of autoimmunity, but how human-associated bacteria impact Th17-driven disease remains elusive. We show that human gut Actinobacterium Eggerthella lenta induces intestinal Th17 activation by lifting inhibition of the Th17

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service