Skip to Content
Merck
All Photos(1)

Key Documents

U4127

Sigma-Aldrich

Urocortin human

≥85% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C204H337N63O64
CAS Number:
Molecular Weight:
4696.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

Assay

≥85% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... UCN(7349)

Amino Acid Sequence

Asp-Asn-Pro-Ser-Leu-Ser-Ile-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Thr-Leu-Leu-Glu-Leu-Ala-Arg-Thr-Gln-Ser-Gln-Arg-Glu-Arg-Ala-Glu-Gln-Asn-Arg-Ile-Ile-Phe-Asp-Ser-Val-NH2

Application

Urocortin human can be used as a test compound for studying the impact of corticotropin-releasing factor (CRF) receptor agonist and antagonist on sleep-wake patterns in conscious rats by using radiotelemetry system.

Biochem/physiol Actions

Binds to CRF receptor types 1, 2α, and 2β; stimulates cAMP accumulation from cells transfected with these receptors. Acts in vitro to release ACTH from rat anterior pituitary cells.
Urocortin is a CRF-related neuropeptide having high affinity for CRF2 receptor possess appetite-suppressing effect. Urocortin along with corticotropin-releasing factor (CRF) binding protein (CRF-BP) enhances the free CRF levels in human brain by displacing hCRF from the binding protein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D P Behan et al.
Brain research, 725(2), 263-267 (1996-07-01)
Here we demonstrate that urocortin, a new mammalian member of the corticotropin-releasing factor (CRF) neuropeptide family has high affinity for both the recombinant human CRF binding protein (CRF-BP) and for a membrane-associated form of the protein solubilized from postmortem human
Masayuki Uchida et al.
Biological & pharmaceutical bulletin, 30(10), 1895-1897 (2007-10-06)
Stress has been known to release corticotropin-releasing factor (CRF) and have an affect on sleep-wake patterns. However, there is no direct evidence of CRF receptor agonist and antagonist on sleep-wake patterns. Therefore, this study aimed to clarify this point by
M Spina et al.
Science (New York, N.Y.), 273(5281), 1561-1564 (1996-09-13)
The neuropeptide corticotropin-releasing factor (CRF) is well known to act on the central nervous system in ways that mimic stress and result in decreases in exploration, increases in sympathetic activity, decreases in parasympathetic outflow, and decreases in appetitive behavior. Urocortin
Hock-Kean Liew et al.
Neuropeptides, 52, 89-96 (2015-06-10)
Urocortin exerts neuroprotective effects in intracerebral hemorrhage (ICH) of rats. For pre-clinical trial, we intended to study the neuroprotective efficacy of human UCN (hUCN)-1, -2 and -3 in treating ICH rats. ICH was induced by infusing bacterial collagenase VII (0.23

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service