Skip to Content
Merck
All Photos(2)

Key Documents

U1752

Sigma-Aldrich

Uridine 5′-monophosphate

≥98%

Synonym(s):

U 5′-P, UMP, Uridylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N2O9P
CAS Number:
Molecular Weight:
324.18
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

natural (organic)

Quality Level

Assay

≥98%

form

powder

solubility

H2O: soluble 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OC1C(O)C(OC1COP(O)(O)=O)N2C=CC(=O)NC2=O

InChI

1S/C9H13N2O9P/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h1-2,4,6-8,13-14H,3H2,(H,10,12,15)(H2,16,17,18)

InChI key

DJJCXFVJDGTHFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Uridine 5′-monophosphate acts as a precursor for all pyrimidine nucleotides.

Application

Uridine 5′-monophosphate has been used as a component in isotonic E3 medium for isotonic reconstitution experiments to study its effects on the basal cell response or epithelial cell migration to the wound in the isotonic medium.

Biochem/physiol Actions

Uridine 5′-monophosphate (UMP) is a nucleotide monomer in RNA. It contains the nucleobase uracil along with the ribose sugar and a phosphate group. UMP dietary supplementation enhances neurotransmitter release and neurite outgrowth and also promotes membrane phosphatide production in adult rats. Oral supplementation of DHA, co-administered with UMP, in adult gerbils shows an increase in dendritic spines and neuronal membrane synthesis. It suggests a strategy for the treatment of cognitive disorders resulting from synapse loss.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaozhen Chen et al.
Journal of pharmaceutical and biomedical analysis, 174, 432-440 (2019-06-21)
A novel, simple, and efficacious analytical method for determining of nucleotides in Anoectochilus roxburghii (Wall.) Lindl (A. roxburghii) was developed. Magnetic dispersive solid-phase micro-extraction (MDSPME) combined with high-performance liquid chromatography was applied for extraction and determination of three nucleotides, such
Regulation of uridylic acid biosynthesis in the cyanobacterium Anabaena variabilis.
Currier TC and Wolk CP
Journal of Bacteriology, 136(2), 682-687 (1978)
Lei Wang et al.
Journal of molecular neuroscience : MN, 27(1), 137-145 (2005-08-02)
Membrane phospholipids like phosphatidylcholine (PC) are required for cellular growth and repair, and specifically for synaptic function. PC synthesis is controlled by cellular levels of its precursor, cytidine-5'-diphosphate choline (CDP-choline), which is produced from cytidine triphosphate (CTP) and phosphocholine. In
Toshimasa Sakamoto et al.
Brain research, 1182, 50-59 (2007-10-24)
Docosahexaenoic acid (DHA), an omega-3 polyunsaturated fatty acid, is an essential component of membrane phosphatides and has been implicated in cognitive functions. Low levels of circulating or brain DHA are associated with various neurocognitive disorders including Alzheimer's disease (AD), while
Chaitanya V Mungi et al.
Life (Basel, Switzerland), 9(3) (2019-07-07)
Polymerization of nucleotides under prebiotically plausible conditions has been a focus of several origins of life studies. Non-activated nucleotides have been shown to undergo polymerization under geothermal conditions when subjected to dry-wet cycles. They do so by a mechanism similar

Protocols

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service