Skip to Content
Merck
All Photos(1)

Key Documents

T9145

Sigma-Aldrich

Thyroglobulin from bovine thyroid

(For Gel Filtration Chromatography)

Synonym(s):

Tg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

bovine thyroid

Quality Level

form

powder

mol wt

~670 kDa

technique(s)

bioconjugation: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... TG(280706)

Looking for similar products? Visit Product Comparison Guide

Application

Thyroglobulin is used for antibody purification and characterization. Thyroglobulin has been used in studies assessing the monitoring of differentiated thyroid cancer as well as to develop an accurate diagnostic tool for patients with autoimmune thyroid diseases.
Often used as a carrier protein for the production of antibodies. Its advantage is the large number of tyrosine residues that can be used for conjugation using the diazo reaction.

Biochem/physiol Actions

Iodine-containing protein that is stored in the thyroid gland. Converted into circulating thyroxines when the thyroid is stimulated by thyroid-stimulating hormone (TSH) from the pituitary. Because its expression is unique to the thyroid, it is a sensitive indicator of recurrence or metastasis of thyroid cancer that has been treated by thyroid ablation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perrine Munier et al.
Annales de biologie clinique, 69(6), 721-725 (2011-11-30)
Thyroglobuline (Tg) is a large molecule of high molecular weight mainly indicated in monitoring differentiated thyroid cancer (DTC), its measurement remains difficult. We report the case of a patient who underwent total thyroidectomy for a poorly differentiated thyroid insular carcinoma.
Andrew N Hoofnagle et al.
Clinical chemistry, 54(11), 1796-1804 (2008-09-20)
Quantification of serum tumor markers plays an important role in determining whether patients treated for cancer require further therapy. Whereas large-scale proteomic efforts aim to identify novel tumor markers to facilitate early detection, optimization of methods for quantifying known tumor
Morgan E Abernathy et al.
NPJ vaccines, 6(1), 126-126 (2021-10-27)
HIV-1 vaccine design aims to develop an immunogen that elicits broadly neutralizing antibodies against a desired epitope, while eliminating responses to off-target regions of HIV-1 Env. We report characterization of Ab1245, an off-target antibody against the Env gp120-gp41 interface, from
Akio Yoshida et al.
The Journal of clinical endocrinology and metabolism, 94(2), 442-448 (2008-12-04)
Pendrin is an apical protein of thyroid follicular cells, responsible for the efflux of iodide into the follicular lumen via an iodide-chloride transport mechanism. It is unknown whether pendrin is recognized by autoantibodies. Our objective was to examine the prevalence
Kookjoo Kim et al.
Acta crystallographica. Section D, Structural biology, 77(Pt 11), 1451-1459 (2021-11-03)
Thyroglobulin is a homodimeric glycoprotein that is essential for the generation of thyroid hormones in vertebrates. Upon secretion into the lumen of follicles in the thyroid gland, tyrosine residues within the protein become iodinated to produce monoiodotyrosine (MIT) and diiodotyrosine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service