Skip to Content
Merck
All Photos(9)

Key Documents

T9026

Sigma-Aldrich

Anti-TUBA4A (TUBA1) Antibody

enhanced validation

mouse monoclonal, DM1A

Synonym(s):

Monoclonal Anti-α-Tubulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

DM1A, monoclonal

mol wt

antigen ~50 kDa

contains

15 mM sodium azide

species reactivity

yeast, mouse, amphibian, human, rat, chicken, fungi, bovine

enhanced validation

independent ( antibodies)
Learn more about Antibody Enhanced Validation

technique(s)

indirect immunofluorescence: 1:500 using cultured chicken fibroblasts
western blot: 1:500 using human or chicken fibroblasts

isotype

IgG1

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBA4A(7277)
mouse ... Tuba1a(22142)
rat ... Tuba1a(64158)

Looking for similar products? Visit Product Comparison Guide

General description

α-tubulin, also called tubulin α 4a (TUBA4A), is mapped to human chromosome 2q35. The gene codes for a member of the α-tubulin family, and contains 448 amino acids. TUBA4A belongs to the subfamily of tubulin, which is the major building block of microtubules. α-subunit of tubulin has molecular weight of 50,000.
Monoclonal Anti-α-Tubulin (mouse IgG1 isotype) is derived from the DM1A hybridoma produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. Purified chick brain microtubules were used as immunogen. The isotype is determined by a double diffusion immunoassay using Mouse Monoclonal Antibody Isotyping Reagents, Product Number ISO2. The product is Protein A purified Monoclonal Anti-α-Tubulin antibody conjugated to fluorescein isothiocyanate, isomer I. It is purified by gel filtration and contains no detectable free FITC. Anti-α-Tubulin FITC antibody, Mouse monoclonal specifically recognizes an epitope in the carboxy terminal part of α-tubulin. It localizes α-tubulin in human, monkey, bovine, chicken, goat, murine, rat, gerbil, hamster, rat kangaroo, amphibia, sea urchin, trypanosome, yeast, fungi and tobacco.

Specificity

Anti alpha-tubulin antibody is specific for α-tubulin in immunoblotting assays and may be used for localization of α-tubulin in cultured cells or tissue sections. The antibody reacts best with chicken fibroblasts.
The antibody is specific for α-tubulin in immunoblotting assays and may be used for localization of α-tubulin in cultured cells or tissue sections. The antibody reacts best with chicken fibroblasts.

Immunogen

microtubules from chicken embryo brain.

Application

Monoclonal Anti-α-Tubulin antibody produced in mouse has been used in western blot analysis and protein expression analysis.
Monoclonal Anti-α-Tubulin antibody produced in mouse has been used in the detection of α-Tubulin:
  • in human osteosarcoma and in breast cancer cell lines by western blotting
  • in HeLa cells by immunofluorescence microscopy
  • by immunostaining
  • immunohistochemical detection in Xenopus embryos

Biochem/physiol Actions

Tubulin α 4a (TUBA4A) mutation is associated with amyotrophic lateral sclerosis (ALS).
α/β-Tubulin is the major building block of microtubules. These intracellular, hollow, cylindrical, filamentous structures are present in virtually all eukaryotic cells. Self-assembly of α/β-tubulin leads to polar microtubular structures built from linearly arranged strings of alternatively α- and β-tubulin pairs pointing in the same direction. Microtubules function as structural and mobile elements in mitosis, intracellular transport, ciliary flagellar motility and generation and maintenance of cell shape. α/β-Tubulin and γ-tubulin are members of the tubulin superfamily of proteins. α/β-Tubulin is a heterodimer which consists of one α-tubulin chain and one β-tubulin chain; each subunit has a molecular weight of 55 kDa and they share considerable homolog.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.
For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MID1 and MID2 are required for Xenopus neural tube closure through the regulation of microtubule organization.
Suzuki M, et al.
Development, dev-048769 (2010)
Analysis of gene function in somatic mammalian cells using small interfering RNAs.
Elbashir SM, et al.
Methods, 26(2), 199-213 (2002)
Telomere maintenance through spatial control of telomeric proteins.
Chen LY
Molecular and Cellular Biology, 27(16), 5898-5909 (2007)
HDAC6 is a microtubule-associated deacetylase.
Hubbert C, et al.
Nature, 417(6887), 455-455 (2002)
CHIP overexpression reduces mutant androgen receptor protein and ameliorates phenotypes of the spinal and bulbar muscular atrophy transgenic mouse model.
Adachi H
The Journal of Neuroscience, 27(19), 5115-5126 (2007)

Articles

Immunofluorescence uses antibody-conjugated fluorescent molecules for protein localization, modification confirmation, and protein complex visualization.

Immunofluorescence uses antibody-conjugated fluorescent molecules for protein localization, modification confirmation, and protein complex visualization.

Immunofluorescence uses antibody-conjugated fluorescent molecules for protein localization, modification confirmation, and protein complex visualization.

Immunofluorescence uses antibody-conjugated fluorescent molecules for protein localization, modification confirmation, and protein complex visualization.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service