Skip to Content
Merck
All Photos(1)

Key Documents

T7915

Sigma-Aldrich

Thioredoxin Reductase from Escherichia coli

ammonium sulfate suspension, >25 units/mg protein (Bradford)

Synonym(s):

NADPH:oxidized thioredoxin oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

biological source

Escherichia coli

Quality Level

form

ammonium sulfate suspension

specific activity

>25 units/mg protein (Bradford)

mol wt

70 kDa

technique(s)

activity assay: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

Escherichia coli K12 ... trxB(949054)

General description

Research area: Cell Signaling
Thioredoxin reductases (TrxRs) belongs to family of selenium-containing pyridine nucleotide-disulphide oxidoreductases.

Application

Thioredoxin Reductase from Escherichia coli can be used in peroxidase-coupled thioredoxin system assay for assessing the peroxidase activitiy of Cys-based thiol peroxidases.Thioredoxin Reductase from Escherichia coli has been used:
  • for determining the enzymatic activity of His6-Ahp1p.
  • to assay C. elegans TRXR using the TXN-dependent activity assay.
  • to investigate the biological reducing systems for organic hydroperoxide resistance R gene (OhrR).
  • for enzymatic targeting of auranofin to test its antimicrobial properties.
  • in thioredoxin reductase activity and peroxiredoxin assay.
  • to study the synergy between broccoli sprout extract and selenium in the upregulation of thioredoxin reductase in human hepatocytes.
Thioredoxin reductase from Escherichia coli has been used in thioredoxin reductase activity and peroxiredoxin assay.
It has also been used to study the synergy between broccoli sprout extract and selenium in the upregulation of thioredoxin reductase in human hepatocytes.

Biochem/physiol Actions

An FAD-containing enzyme involved in the transfer of hydrogen from E. coli thioredoxin to other proteins thus providing a powerful disulfide reductase system.
Thioredoxin reductase is a FAD containing enzyme, which transfers the reducing equivalent from NADPH to the disulphide bond of the enzyme by using FAD moiety within the Cys-Ala-Thr-Cys sequence. It can also reduce Trx-S2 to Trx-(SH)2 by using NADPH.
Thioredoxin reductase (TrxR) is an NADPH-dependent oxidoreductase containing one FAD per subunit that reduces the active site disulfide in oxidised thioredoxin (Trx). The molecular weight of the isozymes from mammalian sources vary between 55-67 kDa as compared with 35 kDa in prokaryotes, plants or yeast. The substrate specificity of the mammalian enzyme is much broader than the prokaryotic enzyme reducing both mammalian and E. coli thioredoxins as well as well as non-disulfide substrates such selenite, lipoic acids, lipid hydroperoxides and hydrogen peroxide.

Unit Definition

One unit will cause an increase in absorbance of 1.0 at 412 nm (when measured in a coupled assay with E. coli thioredoxin and DTNB) per min per mL at pH 7.0 at 25 °C.

Physical form

Suspension in 3.6 M (NH4)2SO4 containing 30 mM potassium phosphate buffer, pH 7.5, and 2 mM EDTA.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synergy between broccoli sprout extract and selenium in the upregulation of thioredoxin reductase in human hepatocytes
Li D, et al.
Food Chemistry, 110(1), 193-198 (2008)
Deletion of Thioredoxin Reductase and Effects of Selenite and Selenate Toxicity in Caenorhabditis elegans
Boehler CJ, et al.
PLoS ONE, 8(8), e71525-e71525 (2013)
Auranofin is an effective agent against clinical isolates of Staphylococcus aureus
Tharmalingam N, et al.
Future medicinal chemistry, 11(12), 1417?1425-1417?1425 (2019)
Thioredoxin and thioredoxin reductase.
A Holmgren et al.
Methods in enzymology, 252, 199-208 (1995-01-01)
SOX2 O-GlcNAcylation alters its protein-protein interactions and genomic occupancy to modulate gene expression in pluripotent cells
Myres SA, et al.
eLife, 5, e10647-e10647 (2016)

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service